Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive control: Human pancreas,
Cellular location: Secreted, Neuroendocrine and endocrine secretory granules.
Chromogranin B is a tyrosine sulfated secretory protein found in a wide variety of peptidergic endocrine cells. Chromogranin functions as a neuroendocrine secretory granule protein which may be the precursor for other biologically active peptides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CgB; Chromogranin B, parathyroid secretory protein; Chromogranin-B; Glucagonoma peptide; secretogranin B; Secretogranin I; Secretogranin-1; Secretogranin-I; SgI
Gene Aliases: CHGB; SCG1
UniProt ID: (Human) P05060, (Rat) O35314
Entrez Gene ID: (Human) 1114, (Rat) 24259
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support