Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
11057-1-AP is a rabbit polyclonal antibody raised against the full-length of human CIRHIN.
Immunogen sequence: RCLISCSKK RQLLLFQFAH HLELWRLGST VATGKNGDTL PLSKNADHLL HLKTKGPENI ICSCISPCGS WIAYSTVSRF FLYRLNYEHD NISLKRVSKM PAFLRSALQI LFSEDSTKLF VASNQGALHI VQLSGGSFKH LHAFQPQSGT VEAMCLLAVS PDGNWLAASG TSAGVHVYNV KQLKLHCTVP AYNFPVTAMA IAPNTNNLVI AHSDQQVFEY SIPDKQYTDW SRTVQKQGFH HLWLQRDTPI THISFHPKRP MHILLHDAYM FCIIDKSLPL PNDKTLLYNP FPPTNESDVI RRRTAHAFKI SKIYKPLLFM DLLDERTLVA VERPLDDIIA QLPPPIKKKK FGT (335-686 aa encoded by BC009348)
CIRH1A encodes a WD40-repeat-containing protein that is localized to the nucleolus. Mutation of this gene causes North American Indian childhood cirrhosis, a severe intrahepatic cholestasis that results in transient neonatal jaundice, and progresses to periportal fibrosis and cirrhosis in childhood and adolescence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cirhin; cirrhosis, autosomal recessive 1A (cirhin); testis expressed gene 292; testis-expressed gene 292; Testis-expressed gene 292 protein; U3 small nucleolar RNA-associated protein 4 homolog; UTP4 small subunit (SSU) processome component; UTP4 small subunit processome component; UTP4, small subunit (SSU) processome component, homolog
Gene Aliases: CIRH1A; CIRHIN; cPERP-E; KIAA1988; NAIC; Teg-292; TEX292; UTP4
UniProt ID: (Mouse) Q8R2N2, (Human) Q969X6
Entrez Gene ID: (Mouse) 21771, (Rat) 291987, (Human) 84916
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support