Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTGEVGSEV HLEINDPNVI SQEEADSPSD SGQGSYETIG PLSEGDSDEE IFVSKKLKNR KVLQDSDSET EDTNASPEKT TYDSAEEENK ENLYAGKNTK IKRIYKTVAD SDESYMEKSL YQENLEAQVK PCLELSLQSG NSTDFTTDRK SSKKHIHDKE GTAGKAKVKS KRRLEKEERK MEKIRQLKKK ETKNQEDDVE QPFNDSGCLL VDKDLFETGL EDENNSPLED EESLESIRAA VKNKVKKHKK KEPSLESGVH SFEEGSELSK GTTRKERKAA RLSKEALKQL HSETQRLIRE SALNLPYHMP ENKTIHDFFK RKPRPTCHGN AMALLKSSKY QSSHHKEIID TANTT (1-354 aa encoded by BC137279)
Xenopus claspin is an essential upstream regulator of checkpoint kinase 1 and triggers a checkpoint arrest of the cell cycle in the presence of DNA templates in Xenopus egg extracts. The human gene appears to be the homolog Xenopus claspin and its function has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Claspin; claspin homolog; hClaspin; MGC131612; MGC131613; MGC131615
Gene Aliases: CLSPN
UniProt ID: (Human) Q9HAW4
Entrez Gene ID: (Human) 63967
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support