Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Plays a major role in tight junction specific obliteration of the intercellular space, through calcium independent cell adhesion activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Claudin-15
Gene Aliases: CLDN15
UniProt ID: (Human) P56746
Entrez Gene ID: (Human) 24146
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support