Search Thermo Fisher Scientific
FIGURE: 1 / 1
Laminins are large heterotrimeric, non-collagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms. Laminin 111 consists of the alpha-1, beta-1, and gamma-1 laminin chains.