Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Cofilin is a widely distributed intracellular actin-modulating protein that binds and depolymerizes filamentous F-actin and inhibits the polymerization of monomeric G-actin in a pH-dependent manner. It is involved in the translocation of actin-cofilin complex from cytoplasm to nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COF2; cofilin 2 (muscle); Cofilin, muscle isoform; Cofilin-2; HGNC:1875; nemaline myopathy type 7
Gene Aliases: CFL2; NEM7
UniProt ID: (Human) Q9Y281, (Mouse) P45591
Entrez Gene ID: (Human) 1073, (Rat) 366624, (Mouse) 12632
Molecular Function:
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support