Product References
Chronic intermittent hypoxia impaired collagen synthesis in mouse genioglossus via ROS accumulation: A transcriptomic analysis.
Respiratory physiology & neurobiology
Zhang MH,Han XX,Lu Y,Deng JJ,Zhang WH,Mao JQ,Mi J,Ding WH,Wu MJ,Yu LM,Liu YH
Wed Feb 01 00:00:00 EST 2023
Developmental growth plate cartilage formation suppressed by artificial light at night via inhibiting BMAL1-driven collagen hydroxylation.
Cell death and differentiation
Chen G,Tang Q,Yu S,Shen Y,Sun J,Peng J,Yin Y,Feng G,Lu X,Mei G,Zhang Y,Wan Q,Zhang L,Chen L
Thu Jun 01 00:00:00 EDT 2023
In vivo rAAV-mediated human TGF-β overexpression reduces perifocal osteoarthritis and improves osteochondral repair in a large animal model at one year.
Osteoarthritis and cartilage
Schrenker S,Cucchiarini M,Goebel L,Oláh T,Venkatesan JK,Schmitt G,Speicher-Mentges S,Maihöfer J,Gao L,Zurakowski D,Menger MD,Laschke MW,Madry H
Sat Apr 01 00:00:00 EDT 2023
High expression of COL6A1 predicts poor prognosis and response to immunotherapy in bladder cancer.
Cell cycle (Georgetown, Tex.)
Zhang X,Liu J,Yang X,Jiao W,Shen C,Zhao X,Wang Y
Wed Mar 01 00:00:00 EST 2023
Exon-Skipping for a Pathogenic COL6A1 Variant in Ullrich Congenital Muscular Dystrophy.
Methods in molecular biology (Clifton, N.J.)
Aguti S,Guirguis F,Bönnemann C,Muntoni F,Bolduc V,Zhou H
Tue Nov 22 00:00:00 EST 2022
Electroacupuncture remodels the extracellular matrix and promotes synaptic plasticity in a mouse model of depression.
Biochemical and biophysical research communications
Feng Y,Zhu G,Chen R,Shi G,Peng M,Zhou Y,Fu W,Zhou P
Thu Oct 20 00:00:00 EDT 2022
Sex-specific phenotypes in the aging mouse heart and consequences for chronic fibrosis.
American journal of physiology. Heart and circulatory physiology
Angelini A,Ortiz-Urbina J,Trial J,Reddy AK,Malovannaya A,Jain A,Entman ML,Taffet GE,Cieslik KA
Mon Aug 01 00:00:00 EDT 2022
Extracellular Matrix-Oriented Proteomic Analysis of Periodontal Ligament Under Mechanical Stress.
Frontiers in physiology
Thant L,Kaku M,Kakihara Y,Mizukoshi M,Kitami M,Arai M,Kitami K,Kobayashi D,Yoshida Y,Maeda T,Saito I,Uoshima K,Saeki M
Sat Jul 16 00:00:00 EDT 2022
Skeletal muscle proteome expression differentiates severity of cancer cachexia in mice and identifies loss of fragile X mental retardation syndrome-related protein 1.
Proteomics
Khamoui AV,Tokmina-Roszyk D,Feresin RG,Fields GB,Visavadiya NP
Sun May 01 00:00:00 EDT 2022
Coaxial structured drug loaded dressing combined with induced stem cell differentiation for enhanced wound healing.
Biomaterials advances
Gao C,Zhang L,Wang J,Cheng Y,Chen Z,Yang R,Zhao G
Tue Mar 01 00:00:00 EST 2022
Coaxial structured drug loaded dressing combined with induced stem cell differentiation for enhanced wound healing.
Biomaterials advances
Gao C,Zhang L,Wang J,Cheng Y,Chen Z,Yang R,Zhao G
Tue Mar 01 00:00:00 EST 2022
Identification of metastasis-associated exoDEPs in colorectal cancer using label-free proteomics.
Translational oncology
Liu X,Li N,Zhang C,Wu X,Zhang S,Dong G,Liu G
Sun May 01 00:00:00 EDT 2022
Identification of metastasis-associated exoDEPs in colorectal cancer using label-free proteomics.
Translational oncology
Liu X,Li N,Zhang C,Wu X,Zhang S,Dong G,Liu G
Sun May 01 00:00:00 EDT 2022
The β8 integrin cytoplasmic domain activates extracellular matrix adhesion to promote brain neurovascular development.
Development (Cambridge, England)
De A,Morales JE,Chen Z,Sebastian S,McCarty JH
Tue Mar 15 00:00:00 EDT 2022
Proteome-wide and matrisome-specific atlas of the human ovary computes fertility biomarker candidates and open the way for precision oncofertility.
Matrix biology : journal of the International Society for Matrix Biology
Ouni E,Nedbal V,Da Pian M,Cao H,Haas KT,Peaucelle A,Van Kerk O,Herinckx G,Marbaix E,Dolmans MM,Tuuri T,Otala M,Amorim CA,Vertommen D
Sun May 01 00:00:00 EDT 2022
Time-Resolved Extracellular Matrix Atlas of the Developing Human Skin Dermis.
Frontiers in cell and developmental biology
Li M,Li X,Liu B,Lv L,Wang W,Gao D,Zhang Q,Jiang J,Chai M,Yun Z,Tan Y,Gong F,Wu Z,Zhu Y,Ma J,Leng L
Wed Dec 15 00:00:00 EST 2021
Gapmer Antisense Oligonucleotides to Selectively Suppress the Mutant Allele in COL6A Genes in Dominant Ullrich Congenital Muscular Dystrophy.
Methods in molecular biology (Clifton, N.J.)
Aguti S,Marrosu E,Muntoni F,Zhou H
Fri Mar 19 00:00:00 EDT 2021
MYCT1 Inhibits the Adhesion and Migration of Laryngeal Cancer Cells Potentially Through Repressing Collagen VI.
Frontiers in oncology
Wang PP,Ding SY,Sun YY,Li YH,Fu WN
Wed Mar 10 00:00:00 EST 2021
Priming integrin α5 promotes human dental pulp stem cells odontogenic differentiation due to extracellular matrix deposition and amplified extracellular matrix-receptor activity.
Journal of cellular physiology
Wang H,Ning T,Song C,Luo X,Xu S,Zhang X,Deng Z,Ma D,Wu B
Thu Aug 01 00:00:00 EDT 2019
Priming integrin α5 promotes human dental pulp stem cells odontogenic differentiation due to extracellular matrix deposition and amplified extracellular matrix-receptor activity.
Journal of cellular physiology
Wang H,Ning T,Song C,Luo X,Xu S,Zhang X,Deng Z,Ma D,Wu B
Thu Aug 01 00:00:00 EDT 2019
Differentiation of diabetic foot ulcer-derived induced pluripotent stem cells reveals distinct cellular and tissue phenotypes.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Kashpur O,Smith A,Gerami-Naini B,Maione AG,Calabrese R,Tellechea A,Theocharidis G,Liang L,Pastar I,Tomic-Canic M,Mooney D,Veves A,Garlick JA
Tue Jan 01 00:00:00 EST 2019
Differentiation of diabetic foot ulcer-derived induced pluripotent stem cells reveals distinct cellular and tissue phenotypes.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Kashpur O,Smith A,Gerami-Naini B,Maione AG,Calabrese R,Tellechea A,Theocharidis G,Liang L,Pastar I,Tomic-Canic M,Mooney D,Veves A,Garlick JA
Tue Jan 01 00:00:00 EST 2019
Differentiation of diabetic foot ulcer-derived induced pluripotent stem cells reveals distinct cellular and tissue phenotypes.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Kashpur O,Smith A,Gerami-Naini B,Maione AG,Calabrese R,Tellechea A,Theocharidis G,Liang L,Pastar I,Tomic-Canic M,Mooney D,Veves A,Garlick JA
Tue Jan 01 00:00:00 EST 2019
Differentiation of diabetic foot ulcer-derived induced pluripotent stem cells reveals distinct cellular and tissue phenotypes.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Kashpur O,Smith A,Gerami-Naini B,Maione AG,Calabrese R,Tellechea A,Theocharidis G,Liang L,Pastar I,Tomic-Canic M,Mooney D,Veves A,Garlick JA
Tue Jan 01 00:00:00 EST 2019
Proteomic analysis of mouse ovaries during the prepubertal stages.
Experimental cell research
Xiong J,Wu M,Zhang Q,Zhang C,Xiong G,Ma L,Lu Z,Wang S
Mon Apr 15 00:00:00 EDT 2019
Differential effect of extracellular matrix derived from papillary and reticular fibroblasts on epidermal development in vitro.
European journal of dermatology : EJD
Janson D,Rietveld M,Mahé C,Saintigny G,El Ghalbzouri A
Thu Jun 01 00:00:00 EDT 2017
Authentication of collagen VI antibodies.
BMC research notes
Endicott J,Holden P,Fitzgerald J
Sat Jul 29 00:00:00 EDT 2017
Authentication of collagen VI antibodies.
BMC research notes
Endicott J,Holden P,Fitzgerald J
Sat Jul 29 00:00:00 EDT 2017
Authentication of collagen VI antibodies.
BMC research notes
Endicott J,Holden P,Fitzgerald J
Sat Jul 29 00:00:00 EDT 2017
Authentication of collagen VI antibodies.
BMC research notes
Endicott J,Holden P,Fitzgerald J
Sat Jul 29 00:00:00 EDT 2017
Twist1-induced activation of human fibroblasts promotes matrix stiffness by upregulating palladin and collagen α1(VI).
Oncogene
García-Palmero I,Torres S,Bartolomé RA,Peláez-García A,Larriba MJ,Lopez-Lucendo M,Peña C,Escudero-Paniagua B,Muñoz A,Casal JI
Thu Oct 06 00:00:00 EDT 2016
Twist1-induced activation of human fibroblasts promotes matrix stiffness by upregulating palladin and collagen α1(VI).
Oncogene
García-Palmero I,Torres S,Bartolomé RA,Peláez-García A,Larriba MJ,Lopez-Lucendo M,Peña C,Escudero-Paniagua B,Muñoz A,Casal JI
Thu Oct 06 00:00:00 EDT 2016
Twist1-induced activation of human fibroblasts promotes matrix stiffness by upregulating palladin and collagen α1(VI).
Oncogene
García-Palmero I,Torres S,Bartolomé RA,Peláez-García A,Larriba MJ,Lopez-Lucendo M,Peña C,Escudero-Paniagua B,Muñoz A,Casal JI
Thu Oct 06 00:00:00 EDT 2016
Twist1-induced activation of human fibroblasts promotes matrix stiffness by upregulating palladin and collagen α1(VI).
Oncogene
García-Palmero I,Torres S,Bartolomé RA,Peláez-García A,Larriba MJ,Lopez-Lucendo M,Peña C,Escudero-Paniagua B,Muñoz A,Casal JI
Thu Oct 06 00:00:00 EDT 2016
Upregulation of COL6A1 is predictive of poor prognosis in clear cell renal cell carcinoma patients.
Oncotarget
Wan F,Wang H,Shen Y,Zhang H,Shi G,Zhu Y,Dai B,Ye D
Tue Sep 29 00:00:00 EDT 2015
Upregulation of COL6A1 is predictive of poor prognosis in clear cell renal cell carcinoma patients.
Oncotarget
Wan F,Wang H,Shen Y,Zhang H,Shi G,Zhu Y,Dai B,Ye D
Tue Sep 29 00:00:00 EDT 2015
Reactive stroma component COL6A1 is upregulated in castration-resistant prostate cancer and promotes tumor growth.
Oncotarget
Zhu YP,Wan FN,Shen YJ,Wang HK,Zhang GM,Ye DW
Wed Jun 10 00:00:00 EDT 2015
Reactive stroma component COL6A1 is upregulated in castration-resistant prostate cancer and promotes tumor growth.
Oncotarget
Zhu YP,Wan FN,Shen YJ,Wang HK,Zhang GM,Ye DW
Wed Jun 10 00:00:00 EDT 2015
A new approach for the study of lung smooth muscle phenotypes and its application in a murine model of allergic airway inflammation.
PloS one
Paez-Cortez J,Krishnan R,Arno A,Aven L,Ram-Mohan S,Patel KR,Lu J,King OD,Ai X,Fine A
Tue Jun 03 00:00:00 EDT 2014
A new approach for the study of lung smooth muscle phenotypes and its application in a murine model of allergic airway inflammation.
PloS one
Paez-Cortez J,Krishnan R,Arno A,Aven L,Ram-Mohan S,Patel KR,Lu J,King OD,Ai X,Fine A
Tue Jun 03 00:00:00 EDT 2014