Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Collagen V Antibodies

            Invitrogen

            Collagen V (soluble) Monoclonal Antibody (1E2-E4/Col5)

            View all (5) Collagen V antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # CSI 006-01-02
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            886.00
            400 µL
             
            Estimated availability date: 18-Jun-2025
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite Collagen V (soluble) Monoclonal Antibody (1E2-E4/Col5)

            Product Details

            CSI 006-01-02

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            Assay-dependent
            -

            Immunohistochemistry (Frozen) (IHC (F))

            Assay-dependent
            -

            ELISA (ELISA)

            1:50
            -
            Product Specifications

            Species Reactivity

            Bovine, Dog, Human, Marsupial, Sheep, Pig, Rabbit

            Host/Isotype

            Mouse / IgG2a, kappa

            Class

            Monoclonal

            Type

            Antibody

            Clone

            1E2-E4/Col5

            Immunogen

            Acid-digested pepsin soluble dog type V collagen.

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1.05 mg/mL

            Purification

            Protein A/G

            Storage buffer

            PBS, pH 7.4

            Contains

            15mM sodium azide

            Storage conditions

            4°C, store in dark

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_1073787

            Product Specific Information

            CSI 006-01-02 is highly specific for type V collagen. It has been shown to have no cross-reactivity with type I, III and VI collagens by ELISA and immunoblotting. There is no evidence for cross-reactivity with other connective tissue proteins (laminin, fibronectin, elastin). CSI 006-01-02 shows reactivity with human, dog, calf, sheep, pig and kangaroo in type V collagens by ELISA and immunoblotting. CSI 006-01-02 does not show reactivity with mouse, rat, guinea pig and chicken type V collagens by ELISA and immunoblotting.

            CSI 006-01-02 has been successfully used in Western blotting, ELISA and Immunohistochemistry procedures. CSI 006-01 has been used succesfully for immunohistology on frozen unfixed sections of human, bovine and dog skin, of foetal bovine cornea, and of new dog tissue associated with a biomaterial implant. If fixation of tissue is required, acetone or ethanol is recommended. CSI 006-01 binds poorly to Collagen V when tested in ELISA with Collagen V coated directly onto the microtiter well.

            NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL

            Target Information

            COL5A1 encodes an alpha chain for one of the lowabundance fibrillar collagens. Fibrillar collagen molecules aretrimers that can be composed of one or more types of alpha chains.Type V collagen is found in tissues containing type I collagen andappears to regulate the assembly of heterotypic fibers composed ofboth type I and type V collagen. COL5A1 product is closelyrelated to type XI collagen and it is possible that the collagenchains of types V and XI constitute a single collagen type withtissue-specific chain combinations. Mutations in this gene areassociated with Ehlers-Danlos syndrome, types I and II.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: AB collagen; alpha-1 type V collagen; COL5; Collagen 5; Collagen alpha-1(V) chain; collagen type 5 alpha 1; collagen, type V, alpha 1; procollagen alpha 1(V); RP11-263F14.1; Type V Collagen

            View more View less

            Gene Aliases: COL5A1; EDSC

            View more View less

            UniProt ID: (Human) P20908

            View more View less

            Entrez Gene ID: (Pig) 397533, (Sheep) 101107777, (Dog) 480684, (Rabbit) 100327268, (Human) 1289

            View more View less

            Function(s)

            integrin binding extracellular matrix structural constituent heparin binding proteoglycan binding metal ion binding platelet-derived growth factor binding extracellular matrix structural protein

            Process(es)

            blood vessel development heart morphogenesis cell adhesion cell migration extracellular matrix organization collagen fibril organization collagen catabolic process collagen biosynthetic process wound healing, spreading of epidermal cells tendon development extracellular fibril organization skin development integrin biosynthetic process eye morphogenesis regulation of cellular component organization negative regulation of endodermal cell differentiation
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-bwv9l:80/100.66.73.191:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0