Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acylation-stimulating protein cleavage product; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a anaphylatoxin; Complement C3; complement component 3; complement component C3a; complement component C3b; epididymis secretory sperm binding protein Li 62p; HSE-MSF; prepro-C3
Gene Aliases: AHUS5; AI255234; ARMD9; ASP; C3; C3a; C3b; CPAMD1; HEL-S-62p; HSE-MSF; Plp
UniProt ID: (Rat) P01026, (Human) P01024, (Mouse) P01027, (Bovine) Q2UVX4
Entrez Gene ID: (Rat) 24232, (Human) 718, (Mouse) 12266, (Bovine) 280677
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support