Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
There is a suggested incubation period of 30-60 minutes at room temperature.
Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acylation-stimulating protein cleavage product; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a anaphylatoxin; Complement C3; complement C3 protein (GPC3) precursor; complement component 3; complement component 3d; complement component C3; complement component C3a; complement component C3b; complement component C3d; epididymis secretory sperm binding protein Li 62p; HSE-MSF; prepro-C3
Gene Aliases: AHUS5; AI255234; ARMD9; ASP; C3; C3a; C3b; C3d; CPAMD1; HEL-S-62p; HSE-MSF; Plp
UniProt ID: (Guinea pig) P12387, (Rat) P01026, (Human) P01024, (Mouse) P01027, (Bovine) Q2UVX4, (Pig) P01025
Entrez Gene ID: (Guinea pig) 100135462, (Dog) 476728, (Goat) 100860826, (Rat) 24232, (Human) 718, (Mouse) 12266, (Bovine) 280677, (Pig) 397072, (Sheep) 101108131
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support