Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
MA1-40162 detects C5a/C5a des-Arg from human samples.
MA1-40162 has been successfully used in ELISA, Immunohistochemsitry (Frozen, Paraffin), Functional Assays and Western blot applications.
The MA1-40162 immunogen is the Neo-epitope on C5a.
The MA1-40162 immunogen is the Neo-epitope on C5a (ISHKDMQLG amino acid 65-73).
Endotoxin <24 EU/mg.
Positive control Human Granulocytes.
General comment: Please notice that under given conditions it is known that C5 can expose epitopes normally only found in the cleaved activation products
The protein encoded by this gene is the fifth component of complement, which plays an important role in inflammatory and cell killing processes. This protein is comprised of alpha and beta polypeptide chains that are linked by a disulfide bridge. An activation peptide, C5a, which is an anaphylatoxin that possesses potent spasmogenic and chemotactic activity, is derived from the alpha polypeptide via cleavage with a convertase. The C5b macromolecular cleavage product can form a complex with the C6 complement component, and this complex is the basis for formation of the membrane attack complex, which includes additional complement components. Mutations in this gene cause complement component 5 deficiency, a disease where patients show a propensity for severe recurrent infections. Defects in this gene have also been linked to a susceptibility to liver fibrosis and to rheumatoid arthritis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C4B1; C4B12; C4B2; C4B3; C4B5; C4F; C5a anaphylatoxin; CH; CO4; Complement C5; CPAMD3; DADB-112B14.11; FLJ17816; FLJ17822; FLJ60561; MGC142298; MGC164979; prepro-C5
Gene Aliases: C5; C5a; C5b; C5D; CPAMD4; ECLZB
UniProt ID: (Human) P01031
Entrez Gene ID: (Human) 727
Molecular Function:
protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support