Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Immunogen sequence: EDCNELPPRR NTEILTGSWS DQTYPEGTQA IYKCRPGYRS LGNVIMVCRK GEWVALNPLR KCQKRPCGHP GDTPFGTFTL TGGNVFEYGV KAVYTCNEGY QLLGEINYRE CDTDGWTNDI PICEVVKCLP VTAPENGKIV SSAMEPDREY HFGQAVRFVC NSGYKIEGDE EMHCSDDGFW SKEKPKCVEI SCKSPDVING SPISQKIIYK ENERFQYKCN MGYEYSERGD AVCTESGWRP LPSCEEKSCD NPYIPNGDYS PLRIKHRTGD EITYQCRNGF YPATRGNTAK CTSTGWIPAP RCTLK
Positive samples: Human plasma, Mouse liver, Mouse plasma, Rat liver; Cellular location: Secreted
Factor H is a serum protein acting as an inhibitor in the complement activation system. I activates the C3/C5 convertase by dissociating its subunits and also acts as a cofactor for the enzyme activity of Factor I.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adrenomedullin binding protein; age-related maculopathy susceptibility 1; beta-1-H-globulin; beta-1H; Complement factor H; complement factor H, isoform b; factor H; factor H-like 1; FHL1 antibody; H factor 1; H factor 1 (complement); H factor 2 (complement); HGNC:4883; MGC88246; RP1-177P10.1
Gene Aliases: AHUS1; AMBP1; ARMD4; ARMS1; CFH; CFHL3; FH; FHL1; HF; HF1; HF2; HUS
UniProt ID: (Human) P08603
Entrez Gene ID: (Human) 3075
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support