Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Proteintech
FIGURE: 1 / 6
Immunogen sequence: LPPRRNTEI LTGSWSDQTY PEGTQAIYKC RPGYRSLGNV IMVCRKGEWV ALNPLRKCQK RPCGHPGDTP FGTFTLTGGN VFEYGVKAVY TCNEGYQLLG EINYRECDTD GWTNDIPICE VVKCLPVTAP ENGKIVSSAM EPDREYHFGQ AVRFVCNSGY KIEGDEEMHC SDDGFWSKEK PKCVEISCKS PDVINGSPIS QKIIYKENER FQYKCNMGYE YSERGDAVCT ESGWRPLPSC EEKSCDNPYI PNGDYSPLRI KHRTGDEITY QCRNGFYPAT RGNTAKCTST GWIPAPRCTL KPCDYPDIKH GGLYHENMRR PYFP (24-346 aa encoded by BC037285)
Factor H is a serum protein acting as an inhibitor in the complement activation system. I activates the C3/C5 convertase by dissociating its subunits and also acts as a cofactor for the enzyme activity of Factor I.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adrenomedullin binding protein; age-related maculopathy susceptibility 1; beta-1-H-globulin; beta-1H; Complement factor H; complement factor H, isoform b; factor H; factor H-like 1; FHL1 antibody; H factor 1; H factor 1 (complement); H factor 2 (complement); HGNC:4883; MGC88246; RP1-177P10.1
Gene Aliases: AHUS1; AMBP1; ARMD4; ARMS1; CFH; CFHL3; FH; FHL1; HF; HF1; HF2; HUS
UniProt ID: (Human) P08603
Entrez Gene ID: (Human) 3075
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support