Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Connexin 30.2 Antibodies

            Invitrogen

            Connexin 30.2 Polyclonal Antibody

            1 Published Figure
            8 References
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite Connexin 30.2 Polyclonal Antibody

            • Antibody Testing Data (5)
            • Published Figures (1)
            Application
            Connexin 30.2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 6

            Connexin 30.2 Antibody (40-7400) in IHC (P)

            Immunohistochemistry analysis of Connexin 30.2 showing staining in the membrane and cytoplasm of paraffin-embedded human heart tissue (right) compared to a negative control without primary antibody (left). To expose target proteins, antigen retrieval was performed using 10mM sodium citrate (pH 6.0), microwaved for 8-15 min. Following antigen retrieval, tissues were blocked in 3% H2O2-methanol for 15 min at ... View More
            View Product

            Connexin 30.2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Connexin 30.2 Antibody in Immunohistochemistry (IHC)
            Connexin 30.2 Antibody in Immunohistochemistry (IHC)
            Connexin 30.2 Antibody in Immunohistochemistry (IHC)
            Connexin 30.2 Antibody in Western Blot (WB)
            Connexin 30.2 Antibody in Immunocytochemistry, Immunohistochemistry (PFA fixed) (ICC/IF, IHC (PFA))

            Product Details

            40-7400

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1 µg/mL
            View 2 publications 2 publications

            Immunohistochemistry (IHC)

            -
            View 3 publications 3 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:10-1:50
            -

            Immunohistochemistry (PFA fixed) (IHC (PFA))

            -
            View 1 publication 1 publication

            Immunocytochemistry (ICC/IF)

            2-4 µg/mL
            View 2 publications 2 publications
            Product Specifications

            Species Reactivity

            Human, Mouse

            Published species

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Synthetic peptide derived from the c-term region of the mouse connexin 30.2 protein
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.25 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.4

            Contains

            0.1% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2533480

            Product Specific Information

            40-7400 was used in the western blot analysis to successfully detect Connexin 30.2 in HeLa cells transfected with Cx30.2 and in immunofluorescence (IHC) analysis to successfully detect Connexin 30.2 in mouse heart sections.

            Target Information

            Gap junctions are responsible for transport of ions and metabolites between adjacent cells. These structures are made of two hemichannels, each formed by six connexin molecules. Connexin 30, coded by Cjb6 gene, is a member of the connexin family and is highly expressed in brain and skin. Immunohistochemically connexin 30 was localized in astrocytes, at gap junctions between these cells and on the astrocyte side of gap junctions between astrocytes and oligodendrocytes. Co-localization with connexin 43 was also observed in this study. Cytoskeleton, especially actin filaments, are important components in the processes of assembly, trafficking and stabilization of connexin 30 gap junctions. Knock-out mice studies have also demonstrated that connexin 30 deficiency impairs renal tubular ATP release and pressure natriuresis highlighting the importance of this molecule in kidney. Further, in vitro studies have also demonstrated that connexin 30 overexpression enhances cell proliferation.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: connexin 30.2; connexin 31.9; Connexin-30.2; Connexin-31.9; Cx30.2; Cx31.9; Gap junction alpha-11 protein; Gap junction chi-1 protein; Gap junction delta-3 protein; gap junction membrane channel protein chi 1; gap junction protein, delta 3, 31.9kDa

            View more View less

            Gene Aliases: Cx30.2; CX31.9; GJA11; GJC1; GJD3

            View more View less

            UniProt ID: (Human) Q8N144, (Mouse) Q91YD1

            View more View less

            Entrez Gene ID: (Human) 125111, (Mouse) 353155

            View more View less

            Function(s)

            ion channel activity gap junction channel activity protein binding gap junction

            Process(es)

            cell communication response to glucose gap junction assembly ion transmembrane transport transmembrane transport
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-fsh6n:80/100.66.78.219:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0