Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: RLYKDYDMPRV VACSVEPCPH TVDCYISR
Highest antigen sequence identity to the following orthologs - mouse 86%, rat 90%.
This gene encodes a transmembrane connexin protein that is a component of gap junctions. Mutations in this gene have been associated with erythrokeratodermia variabilis, progressive symmetric erythrokeratoderma and hearing impairment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: connexin 30.3; Connexin-30.3; Cx30.3; Gap junction beta-4 protein; gap junction protein, beta 4, 30.3kDa
Gene Aliases: CX30.3; EKV; GJB4
UniProt ID: (Human) Q9NTQ9
Entrez Gene ID: (Human) 127534
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support