Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: ERERKHHLKH GPNAPSLYDN LSKKRGGLWW TYLLSLIFKA AVDAGFLYIF HRLYKDYDMP RVVACSVEPC PHTVDCYISR PTEKKVFTYF MVTTAAICIL L; Positive Samples: Mouse liver, Mouse skin; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, gap junction
This gene encodes a transmembrane connexin protein that is a component of gap junctions. Mutations in this gene have been associated with erythrokeratodermia variabilis, progressive symmetric erythrokeratoderma and hearing impairment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: connexin 30.3; Connexin-30.3; Cx30.3; Gap junction beta-4 protein; gap junction membrane channel protein beta 4
Gene Aliases: Cnx30.3; Cx30.3; Cxn-30.3; Gjb-4; Gjb4
UniProt ID: (Mouse) Q02738
Entrez Gene ID: (Rat) 117055, (Mouse) 14621
Molecular Function:
gap junction
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support