Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDWKKLQDLL SGVNQYSTAF GRIWLSVVFV FRVLVYVVAA ERVWGDEQKD
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 79%; Dog: 83%; Horse: 83%; Human: 83%; Mouse: 100%; Pig: 83%; Rabbit: 79%; Rat: 100%; Zebrafish: 92%
Gap junctions are conduits that allow the direct cell-to-cell passage of small cytoplasmic molecules, including ions, metabolic intermediates, and second messengers, and thereby mediate intercellular metabolic and electrical communication. Gap junction channels consist of connexin protein subunits, which are encoded by a multigene family. GJBs (gap-junction proteins or connexins) play crucial functional roles associated with these channels. Defects in GJB3 have been linked to erythrokeratodermia variabilis (EKV) is an autosomal dominant genodermatosis characterized by transient figurate red patches or hyperkeratosis. Mutations in GJB2 have also been associated with genetically derived hearing impairments, including autosomal recessive nonsyndromic deafness.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: connexin 31; Connexin-31; Cx31; Gap junction beta-3 protein; gap junction membrane channel protein beta 3
Gene Aliases: Cnx31; Cx31; Cxn-31; D4Wsu144e; Gjb-3; Gjb3
UniProt ID: (Mouse) P28231
Entrez Gene ID: (Mouse) 14620
Molecular Function:
gap junction
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support