Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 4
This antibody is predicted to react with Monkey, Pig, Goat and Rabbit.
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Connexin-36 (CX36), is a member of the connexin gene family that is expressed predominantly in mammalian neurons. Connexins associate in groups of 6 and are organized radially around a central pore to form connexons. Each gap junction intercellular channel is formed by the conjunction of 2 connexons. Mutations in connexin-encoding genes can lead to functional and developmental abnormalities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: connexin 36; Connexin-36; Cx36; Gap junction alpha-9 protein; Gap junction delta-2 protein; gap junction membrane channel protein alpha 9; gap junction protein, delta 2, 36kDa; GJA10
Gene Aliases: CX36; GJA9; GJD2
UniProt ID: (Human) Q9UKL4, (Rat) O70610
Entrez Gene ID: (Human) 57369, (Rat) 50564
Molecular Function: gap junction
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support