Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Gap junctions were first characterized by electron microscopy as regionally specialized structures on plasma membranes of contacting adherent cells. These structures were shown to consist of cell-to-cell closely packed transmembrane channels. Proteins, called connexins, purified from fractions of enriched gap junctions from different tissues differ. Connexins are designated by their molecular mass. Another system of nomenclature divides gap junction proteins into 2 categories, alpha and beta, according to sequence similarities at the nucleotide and amino acid levels. For example, CX43 is designated alpha-1 gap junction protein, whereas CX32 and CX26 are called beta-1 and beta-2 gap junction proteins, respectively. This nomenclature emphasizes that CX32 and CX26 are more homologous to each other than either of them is to CX43. Connexins have four transmembrane, three intracellular, and two extracellular regions. Different tissues express different connexins, though tissue specificities overlap, and a given tissue or cell can express several different connexins. Developmental regulation of at least some of the connexin genes has been found. Embryo implantation is regulated in part by temporally changing patterns of expression of connexins in the embryo and the maternal decidua.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: connexin 45; Connexin-45; CTC-296K1.4; Cx45; Gap junction alpha-7 protein; gap junction channel protein connexin 45; Gap junction gamma-1 protein; gap junction membrane channel protein alpha 7; gap junction protein, gamma 1, 45kDa
Gene Aliases: C130009G16Rik; Cnx45; CX45; Cxn-45; Gja-7; GJA7; GJC1
UniProt ID: (Human) P36383, (Rat) A4GG66, (Mouse) P28229
Entrez Gene ID: (Human) 10052, (Rat) 266706, (Mouse) 14615
Molecular Function:
gap junction
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support