Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Immunogen sequence: APASRTGSAT SAGTVGEQGR PGTHERPGAK PRAGSEKGSA SSRDGKTTVW
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Human: 100%; Rabbit: 100%
This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Connexin-46.6; Connexin-47; Cx46.6; Cx47; CXG2; Gap junction alpha-12 protein; Gap junction gamma-2 protein; gap junction protein, gamma 2, 47kDa
Gene Aliases: CX46.6; Cx47; GJA12; GJC2; HLD2; LMPH1C; PMLDAR; SPG44
UniProt ID: (Human) Q5T442
Entrez Gene ID: (Human) 57165
Molecular Function:
gap junction
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support