Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody is predicted to react with human, rat and dog based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
The protein encoded by this gene is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: axonal-associated cell adhesion molecule; BIG-2; brain-derived immunoglobulin superfamily molecule; Brain-derived immunoglobulin superfamily protein 2; CNTN4A; Contactin-4; MGC33615; SCA16
Gene Aliases: 9630050B05; Axcam; BIG-2; Cntn4; Kiaa3024
UniProt ID: (Mouse) Q69Z26
Entrez Gene ID: (Mouse) 269784
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support