Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Members of the coronin family, such as CORO1B, are WD repeat-containing actin-binding proteins that regulate cell motility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coronin, actin binding protein, 1B; Coronin-1B; Coronin-2
Gene Aliases: CORO1B; CORONIN-2
UniProt ID: (Human) Q9BR76
Entrez Gene ID: (Human) 57175
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support