Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Exalpha
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Recognizes native and denatured forms of B-Actin (42kDa).
Corticotropin-releasing hormone (CRH; 122560) is a 41-amino acid peptide synthesized in the hypothalamus, and play a role in the neuroregulation, responses to stress and immune challenge. Corticotropin Releasing Factor Receptor 2 (CRHR2) was the receptor of CRH. The sepression of CRHR2 has been reported in various regions of the brain as well as in placenta, umbilical vein, heart, epididymis, gastrointestinal tract, adrenal and skeletal muscle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Corticotropin-releasing factor receptor 2; Corticotropin-releasing hormone receptor 2; CRF-R-2; CRF2 receptor, beta isoform; CRH receptor 2 variant B; CRH-R-2; CRH-R2; Crh2 receptor
Gene Aliases: CRF-RB; CRF2; CRF2R; CRFR2; CRH2R; CRHR2; HM-CRF
UniProt ID: (Human) Q13324
Entrez Gene ID: (Human) 1395
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support