Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VGCVAGDEES YEVFKELFDP IISDRHGGYK PTDKHKTDLN HENLKGGDDL
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-CK; Brain creatine kinase; CK MB; CPK-B; CPK-M; Creatine kinase B chain; Creatine kinase B-type; creatine kinase brain; creatine kinase brain-type; Creatine kinase M chain; Creatine kinase M-type; creatine kinase, brain; creatine kinase, muscle; creatine kinase-B; creatine kinase-M; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; M-CK
Gene Aliases: B-CK; BCK; Ck-3; Ck3; CKB; CKBB; CKM; CKMM; HEL-211; HEL-S-29; M-CK; MCK
UniProt ID: (Human) P12277, (Human) P06732, (Mouse) Q04447, (Mouse) P07310
Entrez Gene ID: (Human) 1152, (Human) 1158, (Mouse) 12709, (Mouse) 12715
Molecular Function:
amino acid kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support