Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This antibody can recognize both CKB and CKM due to the high homology.
Immunogen sequence: MPFSNSHNA LKLRFPAEDE FPDLSAHNNH MAKVLTPELY AELRAKSTPS GFTLDDVIQT GVDNPGHPYI MTVGCVAGDE ESYEVFKDLF DPIIEDRHGG YKPSDEHKTD LNPDNLQGGD DLDPNYVLSS RVRTGRSIRG FCLPPHCSRG ERRAIEKLAV EALSSLDGDL AGRYYALKSM TEAEQQQLID DHFLFDKPVS PLLLASGMAR DWPDARGIWH NDNKTFLVWV NEEDHLRVIS MQKGGNMKEV FTRFCTGLTQ IETLFKSKDY EFMWNPHLGY ILTCPSNLGT GLRAGVHIKL PNLGKHEKFS EVLKRLRLQK RGTGGVDTAA VGGVFDVSNA DRLGFSEVEL VQMVVDGVKL LIEMEQRLEQ GQAIDDLMPA QK (1-381 aa encoded by BC001190)
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-CK; Brain creatine kinase; CPK-B; Creatine kinase B chain; Creatine kinase B-type; creatine kinase brain; creatine kinase brain-type; creatine kinase, brain; creatine kinase-B; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29
Gene Aliases: B-CK; BCK; Ck-3; Ck3; CKB; CKBB; Ckbr; HEL-211; HEL-S-29
UniProt ID: (Human) P12277, (Rat) P07335, (Mouse) Q04447
Entrez Gene ID: (Human) 1152, (Rat) 24264, (Mouse) 12709
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support