Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total CtBP1/2.
This gene encodes a protein that binds to the C-terminus of adenovirus E1A proteins. This phosphoprotein is a transcriptional repressor and may play a role during cellular proliferation. This protein and the product of a second closely related gene, CTBP2, can dimerize. Both proteins can also interact with a polycomb group protein complex which participates in regulation of gene expression during development. Alternative splicing of transcripts from this gene results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 50 kDa BFA-dependent ADP-ribosylation substrate; 50-kDa BFA-induced ADP-ribosylated substrate; BARS-50; brefeldin A-ADP-riboslyated substrate; brefeldin A-ribosylated substrate; C-terminal-binding protein 1; C-terminal-binding protein 2; C-terminal-binding protein 3; C-terminus binding protein 3/brefeldin A (BFA) adenosine diphosphate-ribosylated substrate; CtBP1; CtBP2; CtBP3; gene trap ROSA b-geo 6; ribeye
Gene Aliases: 50-kDaBFA-inducedADP-ribosylatedsubstrate; AA407280; BARS; BARS-50; CTBP; CTBP1; CtBP1-L; CtBP1-S; CTBP2; Ctbp3; CtBP3/BARS; D4S115h; D5H4S115; D5H4S115E; D7Ertd45e; Gtrgeo6; Ribeye
UniProt ID: (Human) Q13363, (Human) P56545, (Rat) Q9Z2F5, (Mouse) O88712, (Rat) Q9EQH5, (Mouse) P56546
Entrez Gene ID: (Human) 1487, (Human) 1488, (Rat) 29382, (Mouse) 13016, (Rat) 81717, (Mouse) 13017
Molecular Function: transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support