Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Positive Control: Rat testis tissue lysate, Hela cell lysate, NIH/3T3 cell lysate, rat brain tissue lysate, MCF-7 cell lysate, A431 cell lysate, A549, MCF-7, PANC-1, mouse testis tissue, human breast carcinoma tissue, mouse kidney tissue.
Subcellular Location: Nucleus, synapse.
This gene produces alternative transcripts encoding two distinct proteins. One protein is a transcriptional repressor, while the other isoform is a major component of specialized synapses known as synaptic ribbons. Both proteins contain a NAD+ binding domain similar to NAD+-dependent 2-hydroxyacid dehydrogenases. A portion of the 3' untranslated region was used to map this gene to chromosome 21q21. 3; however, it was noted that similar loci elsewhere in the genome are likely. Blast analysis shows that this gene is present on chromosome 10.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-terminal-binding protein 2; CtBP2; gene trap ROSA b-geo 6; ribeye; RP11-59C5.2
Gene Aliases: AA407280; CTBP2; D7Ertd45e; Gtrgeo6; Ribeye
UniProt ID: (Human) P56545, (Rat) Q9EQH5, (Mouse) P56546
Entrez Gene ID: (Human) 1488, (Rat) 81717, (Mouse) 13017
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support