Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Rockland
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Store vial at 4° C prior to restoration. For extended storage aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This Cybr antibody is directed against mouse Cybr. A BLAST analysis was used to suggest cross-reactivity with Cybr from mouse and rat based on a 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.
The protein encoded by this gene contains 2 leucine zipper domains and a putative C-terminal nuclear targeting signal, but does not have any hydrophobic regions. This protein is expressed weakly in resting NK and T cells. The encoded protein modulates the activation of ARF genes by CYTH1. This protein interacts with CYTH1 and SNX27 proteins and may act to sequester CYTH1 protein in the cytoplasm.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CASP; Cbp HE; CYBR; Cytohesin binder and regulator; cytohesin binding protein; cytohesin binding protein HE; cytohesin-1 interacting protein; Cytohesin-associated scaffolding protein; Cytohesin-binding protein HE; Cytohesin-interacting protein; Pleckstrin homology Sec7 and coiled-coil domain-binding protein; Pleckstrin homology Sec7 and coiled-coil domains-binding protein; pleckstrin homology, Sec7 and coiled-coil domains, binding protein; pleckstrin homology, Sec7 and coiled/coil domains, binding protein; Sec7; tamalin
Gene Aliases: A130053M09Rik; AI462064; B3-1; C80816; CASP; Cbp; CYBR; CYTHIP; CYTIP; HE; PSCDBP
UniProt ID: (Human) O60759, (Rat) Q5I0L6, (Mouse) Q91VY6
Entrez Gene ID: (Human) 9595, (Rat) 311047, (Mouse) 227929
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support