Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Rockland
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Antiserum will specifically react with a 60 kDa Cyclin A protein from human, rat and mouse tissue. No reaction was observed against other related cyclins. Cross reactivity with Cyclin A from other species may also occur.
The critical role that the family of regulatory proteins known as cyclins play in eukaryotic cell cycle regulation is well established. The best-characterized cyclin complex is the mitotic cyclin B/Cdc2 p34 kinase, the active component of maturing promoting factor. Cyclin A accumulates prior to cyclin B in the cell cycle, appears to be involved in control of S phase and has been shown to associate with cyclin-dependent kinase-2 (Cdk2). In addition, cyclin A has been implicated in cell transformation and is found in complexes with E1A, transcription factors DRTF1 and E2F and retinoblastoma protein, p110. A second form of cyclin A, named cyclin A1 because of its high sequence homology to Xenopus cyclin A1, is most highly expressed in germ cells. It has been proposed that cyclin A1 can associate with Cdk2, p39 and Cdc2 p34.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cyclin A; Cyclin A2 (Cyclin A); Cyclin-A; Cyclin-A1; Cyclin-A2; CyclinA; CyclinA1; testicular tissue protein Li 34
Gene Aliases: AA408589; Ccn-1; CCN1; CCNA; CCNA1; CCNA2; CT146; Cyca; CycA2
UniProt ID: (Human) P78396, (Human) P20248, (Rat) Q6AY13, (Mouse) Q61456, (Mouse) P51943
Entrez Gene ID: (Human) 8900, (Human) 890, (Rat) 295052, (Mouse) 12427, (Rat) 114494, (Mouse) 12428
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support