Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Cyclin B2 is a member of the cyclin family, specifically the B-type cyclins. The B-type cyclins, B1 and B2, associate with p34cdc2 and are essential components of the cell cycle regulatory machinery. B1 and B2 differ in their subcellular localization. Cyclin B1 co-localizes with microtubules, whereas cyclin B2 is primarily associated with the Golgi region. Cyclin B2 also binds to transforming growth factor beta RII and thus cyclin B2/cdc2 may play a key role in transforming growth factor beta-mediated cell cycle control.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-B2; G2/mitotic-specific cyclin B2; G2/mitotic-specific cyclin-B2
Gene Aliases: CCNB2; CycB2; cyclinb2; HsT17299; XELAEV_18020679mg
UniProt ID: (Human) O95067, (Mouse) P30276, (Xenopus) P13351
Entrez Gene ID: (Human) 9133, (Rat) 363088, (Mouse) 12442, (Xenopus) 397743
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support