Product References
CDK/cyclin dependencies define extreme cancer cell-cycle heterogeneity and collateral vulnerabilities.
Cell reports
Knudsen ES,Kumarasamy V,Nambiar R,Pearson JD,Vail P,Rosenheck H,Wang J,Eng K,Bremner R,Schramek D,Rubin SM,Welm AL,Witkiewicz AK
MA5-12707 was used in Immunoprecipitation to define multiple mechanisms for G1/S progression in retinoblastoma-proficient models, which are cyclin-dependent kinase (CDK) 4/6 independent and elicit resistance to FDA-approved inhibitors.
Tue Mar 01 00:00:00 UTC 2022
CDK/cyclin dependencies define extreme cancer cell-cycle heterogeneity and collateral vulnerabilities.
Cell reports
Knudsen ES,Kumarasamy V,Nambiar R,Pearson JD,Vail P,Rosenheck H,Wang J,Eng K,Bremner R,Schramek D,Rubin SM,Welm AL,Witkiewicz AK
MA5-12707 was used in Immunoprecipitation to define multiple mechanisms for G1/S progression in retinoblastoma-proficient models, which are cyclin-dependent kinase (CDK) 4/6 independent and elicit resistance to FDA-approved inhibitors.
Tue Mar 01 00:00:00 UTC 2022
CDK/cyclin dependencies define extreme cancer cell-cycle heterogeneity and collateral vulnerabilities.
Cell reports
Knudsen ES,Kumarasamy V,Nambiar R,Pearson JD,Vail P,Rosenheck H,Wang J,Eng K,Bremner R,Schramek D,Rubin SM,Welm AL,Witkiewicz AK
MA5-12707 was used in Immunoprecipitation to define multiple mechanisms for G1/S progression in retinoblastoma-proficient models, which are cyclin-dependent kinase (CDK) 4/6 independent and elicit resistance to FDA-approved inhibitors.
Tue Mar 01 00:00:00 UTC 2022
p27 allosterically activates cyclin-dependent kinase 4 and antagonizes palbociclib inhibition.
Science (New York, N.Y.)
Guiley KZ,Stevenson JW,Lou K,Barkovich KJ,Kumarasamy V,Wijeratne TU,Bunch KL,Tripathi S,Knudsen ES,Witkiewicz AK,Shokat KM,Rubin SM
MA5-12707 was used in Immunoprecipitation to investigate the mechanism of p27's association with active CDK4.
Fri Dec 13 00:00:00 UTC 2019
CDK4 T172 phosphorylation is central in a CDK7-dependent bidirectional CDK4/CDK2 interplay mediated by p21 phosphorylation at the restriction point.
PLoS genetics
Bisteau X,Paternot S,Colleoni B,Ecker K,Coulonval K,De Groote P,Declercq W,Hengst L,Roger PP
MA5-12707 was used in immunoprecipitation to study the roles CDK7 and p21 in regulating the T172 phosphorylation of CDK4 at the restriction point
Wed May 01 00:00:00 UTC 2013
Aurora B kinase phosphorylates and instigates degradation of p53.
Proceedings of the National Academy of Sciences of the United States of America
Gully CP,Velazquez-Torres G,Shin JH,Fuentes-Mattei E,Wang E,Carlock C,Chen J,Rothenberg D,Adams HP,Choi HH,Guma S,Phan L,Chou PC,Su CH,Zhang F,Chen JS,Yang TY,Yeung SC,Lee MH
MA5-12707 was used in western blot to study the degradation of p53 and the accelerating role of phosphorylation by Aurora B kinase
Tue Jun 12 00:00:00 UTC 2012
Orphan receptor small heterodimer partner suppresses tumorigenesis by modulating cyclin D1 expression and cellular proliferation.
Hepatology (Baltimore, Md.)
Zhang Y,Xu P,Park K,Choi Y,Moore DD,Wang L
MA5-12707 was used in immunoprecipitation to study the mechanism by which the nuclear receptor family member SHP suppresses hepatic carcinogenesis
Tue Jul 01 00:00:00 UTC 2008
Cyclic AMP inhibits the proliferation of thyroid carcinoma cell lines through regulation of CDK4 phosphorylation.
Molecular biology of the cell
Rocha AS,Paternot S,Coulonval K,Dumont JE,Soares P,Roger PP
MA5-12707 was used in immunoprecipitation to study the role of CDK4 phosphorylation in the mechanism by which cAMP inhibits thyroid carcinoma cell proliferation
Sat Nov 01 00:00:00 UTC 2008
Molecular characterization of spontaneous mesenchymal stem cell transformation.
PloS one
Rubio D,Garcia S,Paz MF,De la Cueva T,Lopez-Fernandez LA,Lloyd AC,Garcia-Castro J,Bernad A
MA5-12707 was used in western blot to investigate the molecular mechanisms of spontaneous mesenchymal stem cell transformation
Wed Jan 02 00:00:00 UTC 2008
Cyclin D1 antagonizes BRCA1 repression of estrogen receptor alpha activity.
Cancer research
Wang C,Fan S,Li Z,Fu M,Rao M,Ma Y,Lisanti MP,Albanese C,Katzenellenbogen BS,Kushner PJ,Weber B,Rosen EM,Pestell RG
MA5-12707 was used in western blot to study the opposing effect of cyclin D1 and estrogen receptor on BRCA1 repression
Mon Aug 01 00:00:00 UTC 2005
p21Cip1 and p27Kip1 induce distinct cell cycle effects and differentiation programs in myeloid leukemia cells.
The Journal of biological chemistry
Muñoz-Alonso MJ,Acosta JC,Richard C,Delgado MD,Sedivy J,León J
MA5-12707 was used in immunoprecipitation to study the role of p21Cip1 and p27Kip1 in the cell cycle and differentiation programs of myeloid leukemia cells
Fri May 06 00:00:00 UTC 2005
Cyclin C/cdk3 promotes Rb-dependent G0 exit.
Cell
Ren S,Rollins BJ
MA5-12707 was used in immunoprecipitation to study the role of cyclin C/cdk3 in promoting Rb-dependent G0 exit
Fri Apr 16 00:00:00 UTC 2004
Senescence-associated changes in respiration and oxidative phosphorylation in primary human fibroblasts.
The Biochemical journal
Hutter E,Renner K,Pfister G,Stöckl P,Jansen-Dürr P,Gnaiger E
MA5-12707 was used in western blot to study senescence-related changes in respiration and oxidative phosphorylation in primary human fibroblasts
Tue Jun 15 00:00:00 UTC 2004
Differential effects of p21(WAF1/CIP1) deficiency on MMTV-ras and MMTV-myc mammary tumor properties.
Cancer research
Bearss DJ,Lee RJ,Troyer DA,Pestell RG,Windle JJ
MA5-12707 was used in immunoprecipitation to study the role of the CDK inhibitor p21(WAF1/CIP1) in tumor suppression in MMTV-ras and MMTV-myc transgenic mice
Mon Apr 01 00:00:00 UTC 2002
Reconstitution of cyclin D1-associated kinase activity drives terminally differentiated cells into the cell cycle.
Molecular and cellular biology
Latella L,Sacco A,Pajalunga D,Tiainen M,Macera D,D'Angelo M,Felici A,Sacchi A,Crescenzi M
MA5-12707 was used in immunoprecipitation to study the ability of restoring cyclin D1-associated kinase activity to drive terminally differentiated myotubes to enter the cell cycle
Wed Aug 01 00:00:00 UTC 2001
Metabolic stabilization of p27 in senescent fibroblasts correlates with reduced expression of the F-box protein Skp2.
Experimental gerontology
Wagner M,Hampel B,Hütter E,Pfister G,Krek W,Zwerschke W,Jansen-Dürr P
MA5-12707 was used in immunoprecipitation to study the correlation of p27(KIP1) accumulation in senescent fibroblasts with reduced levels of the F-box protein Skp2
Sat Dec 01 00:00:00 UTC 2001
Dominant negative c-jun inhibits activation of the cyclin D1 and cyclin E kinase complexes.
Molecular biology of the cell
Hennigan RF,Stambrook PJ
MA5-12707 was used in immunoprecipitation to investigate the effect of the dominant negative c-jun on the activation of cyclin D1 and cyclin E kinase complexes
Wed Aug 01 00:00:00 UTC 2001
Dominant negative c-jun inhibits activation of the cyclin D1 and cyclin E kinase complexes.
Molecular biology of the cell
Hennigan RF,Stambrook PJ
MA5-12707 was used in immunoprecipitation to investigate the effect of the dominant negative c-jun on the activation of cyclin D1 and cyclin E kinase complexes
Wed Aug 01 00:00:00 UTC 2001
Loss of p27Kip1 from cyclin E/cyclin-dependent kinase (CDK) 2 but not from cyclin D1/CDK4 complexes in cells transformed by polyamine biosynthetic enzymes.
Cancer research
Ravanko K,Järvinen K,Paasinen-Sohns A,Hölttä E
MA5-12707 was used in immunoprecipitation to study the role of p27Kip1 downregulation in tumorigenic transformation induced by polyamine biosynthetic enzyme overexpression
Fri Sep 15 00:00:00 UTC 2000
Progesterone inhibits estrogen-induced cyclin D1 and cdk4 nuclear translocation, cyclin E- and cyclin A-cdk2 kinase activation, and cell proliferation in uterine epithelial cells in mice.
Molecular and cellular biology
Tong W,Pollard JW
MA5-12707 was used in immunohistochemistry to study the molecular mechanisms underlying the effects of female sex hormones on the proliferation of murine uterine epithelial cells
Mon Mar 01 00:00:00 UTC 1999
Inhibition of basal and mitogen-stimulated pancreatic cancer cell growth by cyclin D1 antisense is associated with loss of tumorigenicity and potentiation of cytotoxicity to cisplatinum.
The Journal of clinical investigation
Kornmann M,Arber N,Korc M
MA5-12707 was used in immunoprecipitation to study the effect of cyclin D1 knockdown on the tumorigenicity and cisplantin-sensitivity of PANC-1 human pancreatic cancer cells
Thu Jan 15 00:00:00 UTC 1998
Angiotensin II activation of cyclin D1-dependent kinase activity.
The Journal of biological chemistry
Watanabe G,Lee RJ,Albanese C,Rainey WE,Batlle D,Pestell RG
MA5-12707 was used in immunoprecipitation to investigate the effect of angiotensin II on cyclin D1 expression and cyclin D1-dependent kinase activity in H295R cells
Fri Sep 13 00:00:00 UTC 1996