Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This gene encodes WW domain-containing binding protein 4. The WW domain represents a small and compact globular structure that interacts with proline-rich ligands. This encoded protein is a general spliceosomal protein that may play a role in cross-intron bridging of U1 and U2 snRNPs in the spliceosomal complex A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G1/S-specific cyclin-E2
Gene Aliases: CCNE2; CYCE2
UniProt ID: (Human) O96020, (Mouse) Q9Z238
Entrez Gene ID: (Human) 9134, (Mouse) 12448, (Rat) 362485
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support