Product References
Mechanical Study of Jian-Gan-Xiao-Zhi Decoction on Nonalcoholic Fatty Liver Disease Based on Integrated Network Pharmacology and Untargeted Metabolomics.
Evidence-based complementary and alternative medicine : eCAM
Cao YJ,Li HZ,Zhao J,Sun YM,Jin XW,Lv SQ,Luo JY,Fang XX,Wen WB,Liao JB
45-6100 was used in Western Blotting to investigate the mechanisms by which Jian-Gan-Xiao-Zhi decoction improve nonalcoholic fatty liver disease.
Tue Jul 19 00:00:00 EDT 2022
Impaired disassembly of the axon initial segment restricts mitochondrial entry into damaged axons.
The EMBO journal
Kiryu-Seo S,Matsushita R,Tashiro Y,Yoshimura T,Iguchi Y,Katsuno M,Takahashi R,Kiyama H
Published figure using Cytochrome C monoclonal antibody (Product # 45-6100) in Immunohistochemistry
Mon Oct 17 00:00:00 EDT 2022
Axonal injury alters the extracellular glial environment of the axon initial segment and allows substantial mitochondrial influx into axon initial segment.
The Journal of comparative neurology
Tamada H,Kiryu-Seo S,Sawada S,Kiyama H
45-6100 was used in Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Western Blot to conclude that axonal injury changes the extracellular glial environment surrounding the axon initial segment (AIS) and intracellular mitochondrial localization in the AIS.
Mon Nov 01 00:00:00 EDT 2021
16-Hydroxycleroda-3,13-dien-15,16-olide Induces Apoptosis in Human Bladder Cancer Cells through Cell Cycle Arrest, Mitochondria ROS Overproduction, and Inactivation of EGFR-Related Signalling Pathways.
Molecules (Basel, Switzerland)
Chen YC,Wang PY,Huang BM,Chen YJ,Lee WC,Chen YC
Published figure using Cytochrome C monoclonal antibody (Product # 45-6100) in Western Blot
Sun Aug 30 00:00:00 EDT 2020
16-Hydroxycleroda-3,13-dien-15,16-olide Induces Apoptosis in Human Bladder Cancer Cells through Cell Cycle Arrest, Mitochondria ROS Overproduction, and Inactivation of EGFR-Related Signalling Pathways.
Molecules (Basel, Switzerland)
Chen YC,Wang PY,Huang BM,Chen YJ,Lee WC,Chen YC
Published figure using Cytochrome C monoclonal antibody (Product # 45-6100) in Western Blot
Sun Aug 30 00:00:00 EDT 2020
MicroRNA-325-3p Facilitates Immune Escape of Mycobacterium tuberculosis through Targeting LNX1 via NEK6 Accumulation to Promote Anti-Apoptotic STAT3 Signaling.
mBio
Fu B,Xue W,Zhang H,Zhang R,Feldman K,Zhao Q,Zhang S,Shi L,Pavani KC,Nian W,Lin X,Wu H
45-6100 was used in Western Blotting to contribution to the understanding of the immune escape of M. tuberculosis, and they provide a theoretical basis for the development of therapeutic approaches for drug-resistant TB.
Tue Jun 02 00:00:00 EDT 2020
DNA methylation and histone deacetylation regulating insulin sensitivity due to chronic cold exposure.
Cryobiology
Wang X,Wang L,Sun Y,Li R,Deng J,Deng J
456100 was used in immunohistochemistry and western blot to assess the causal relationship between chronic cold exposure and insulin resistance and the mechanisms of how DNA methylation and histone deacetylation regulate cold-reduced insulin resistance
Wed Feb 01 00:00:00 EST 2017
A versatile tandem RNA isolation procedure to capture in vivo formed mRNA-protein complexes.
Methods (San Diego, Calif.)
Matia-González AM,Iadevaia V,Gerber AP
45-6100 was used in Western Blotting to escribe a tandem RNA isolation procedure (TRIP) that enables purification of in vivo formed messenger ribonucleoprotein (mRNP) complexes.
Sat Apr 15 00:00:00 EDT 2017
Three-dimensional analysis of somatic mitochondrial dynamics in fission-deficient injured motor neurons using FIB/SEM.
The Journal of comparative neurology
Tamada H,Kiryu-Seo S,Hosokawa H,Ohta K,Ishihara N,Nomura M,Mihara K,Nakamura KI,Kiyama H
45-6100 was used in Flow Cytometry, Immunocytochemistry, Western Blot, Immunohistochemistry to demonstrate both 3D images of whole organelle and the intra-organellar structure simultaneously.
Tue Aug 01 00:00:00 EDT 2017
A ketogenic diet accelerates neurodegeneration in mice with induced mitochondrial DNA toxicity in the forebrain.
Neurobiology of aging
Lauritzen KH,Hasan-Olive MM,Regnell CE,Kleppa L,Scheibye-Knudsen M,Gjedde A,Klungland A,Bohr VA,Storm-Mathisen J,Bergersen LH
45-6100 was used in immunohistochemistry - paraffin section to test if the severe neurodegeneration observed in mutUNG1-expressing mice is prevented by a ketogenic diet
Thu Dec 01 00:00:00 EST 2016
Conserved mRNA-binding proteomes in eukaryotic organisms.
Nature structural & molecular biology
Matia-González AM,Laing EE,Gerber AP
Published figure using Cytochrome C monoclonal antibody (Product # 45-6100) in Western Blot
Tue Dec 01 00:00:00 EST 2015
Evolutionarily conserved intercalated disc protein Tmem65 regulates cardiac conduction and connexin 43 function.
Nature communications
Sharma P,Abbasi C,Lazic S,Teng ACT,Wang D,Dubois N,Ignatchenko V,Wong V,Liu J,Araki T,Tiburcy M,Ackerley C,Zimmermann WH,Hamilton R,Sun Y,Liu PP,Keller G,Stagljar I,Scott IC,Kislinger T,Gramolini AO
45-6100 was used in western blot to investigate the impact of Tmem65 on heart development
Fri Sep 25 00:00:00 EDT 2015
Specific acetylation of p53 by HDAC inhibition prevents DNA damage-induced apoptosis in neurons.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Brochier C,Dennis G,Rivieccio MA,McLaughlin K,Coppola G,Ratan RR,Langley B
45-6100 was used in western blot to investigate the effect of p53 acetylation on DNA damage-induced neuronal apoptosis.
Wed May 15 00:00:00 EDT 2013
Photothermal confocal spectromicroscopy of multiple cellular chromophores and fluorophores.
Biophysical journal
Nedosekin DA,Galanzha EI,Ayyadevara S,Shmookler Reis RJ,Zharov VP
Published figure using Cytochrome C monoclonal antibody (Product # 45-6100) in Immunofluorescence
Wed Feb 08 00:00:00 EST 2012
Arrangement of electron transport chain components in bovine mitochondrial supercomplex I1III2IV1.
The EMBO journal
Althoff T,Mills DJ,Popot JL,Kühlbrandt W
Published figure using Cytochrome C monoclonal antibody (Product # 45-6100) in Western Blot
Fri Sep 09 00:00:00 EDT 2011
Mitochondrial DNA toxicity compromises mitochondrial dynamics and induces hippocampal antioxidant defenses.
DNA repair
Lauritzen KH,Cheng C,Wiksen H,Bergersen LH,Klungland A
45-6100 was used in immunohistochemistry - paraffin section to investigate the effects of mitochondrial DNA damage on hippocampal neurons.
Fri Jun 10 00:00:00 EDT 2011
Analysis of mouse models of cytochrome c oxidase deficiency owing to mutations in Sco2.
Human molecular genetics
Yang H,Brosel S,Acin-Perez R,Slavkovich V,Nishino I,Khan R,Goldberg IJ,Graziano J,Manfredi G,Schon EA
Published figure using Cytochrome C monoclonal antibody (Product # 45-6100) in Western Blot
Fri Jan 01 00:00:00 EST 2010