Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Description: A second major function for cytochrome c is its role as an intermediate in apoptosis, both through its association with inositol-3-phosphate receptor (IP3 receptor) which acts as an ER calcium release channel, and its association with apoptotic protease activating factor 1 (Apaf-1), resulting in the formation of the apoptosome protein complex which then recruits and activates pro-caspase 9 (Apaf-3).
This antibody recognises cytochrome c, originally termed Apaf-2, an evolutionarily conserved protein which acts as an essential mitochondrial electron carrier between respirable substrates and oxygen, during aerobic energy production. Clone 2G8 is reported to block the role of cytochrome c as a co-factor in the activation of caspase 9.
Applications Tested: ELISA, Flow Cytometry (Intracellular Staining), Functional Studies (Blocking).
Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CRF; Cytochrome c; hc1
Gene Aliases: CYC; CYCS; HCS; THC4
UniProt ID: (Human) P99999
Entrez Gene ID: (Human) 54205
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support