Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total Cytochrome P450 2C8/9/18/19.
CYP2C9 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. CYP2C9 localizes to the endoplasmic reticulum and its expression is induced by rifampin. The enzyme is known to metabolize many xenobiotics, including phenytoin, tolbutamide, ibuprofen and S-warfarin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (R)-limonene 6-monooxygenase; (S)-limonene 6-monooxygenase; (S)-limonene 7-monooxygenase; (S)-mephenytoin hydroxylase associated cytochrome P450; 4-nitrophenol 2-hydroxylase; Cholesterol 25-hydroxylase; CYPIIC17; CYPIIC18; CYPIIC19; CYPIIC8; CYPIIC9; CYPIIE1; cytochrome P-450 II C; cytochrome P-450 S-mephenytoin 4-hydroxylase; Cytochrome P-450MP; Cytochrome P450 2C18; Cytochrome P450 2C19; Cytochrome P450 2C8; Cytochrome P450 2C9; Cytochrome P450 2E1; Cytochrome P450 form 1; Cytochrome P450 IIC2; Cytochrome P450 MP-12; Cytochrome P450 MP-20; Cytochrome P450 MP-4; Cytochrome P450 MP-8; Cytochrome P450 PB-1; cytochrome P450, 2e1, ethanol inducible; cytochrome P450, family 2, subfamily C, polypeptide 18; cytochrome P450, family 2, subfamily C, polypeptide 19; cytochrome P450, family 2, subfamily C, polypeptide 8; cytochrome P450, family 2, subfamily C, polypeptide 9; cytochrome P450, family 2, subfamily E, polypeptide 1; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 17; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 18; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 19; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 8; cytochrome P450, subfamily IIE (ethanol-inducible), polypeptide 1; Cytochrome P450-11A; Cytochrome P450-254C; Cytochrome P450-6b/29c; Cytochrome P450-ALC; Cytochrome P450-J; Fenbendazole monooxygenase (4'-hydroxylating); flavoprotein-linked monooxygenase; mephenytoin 4'-hydroxylase; Mephenytoin 4-hydroxylase; microsomal monooxygenase; P450 form 1; S-mephenytoin 4-hydroxylase; unspecific monooxygenase; xenobiotic monooxygenase
Gene Aliases: CPC8; CPC9; CPCI; CPCJ; CPE1; CYP2C; CYP2C10; CYP2C17; CYP2C18; CYP2C19; CYP2C8; CYP2C9; CYP2E; Cyp2e-1; CYP2E1; CYPIIC17; CYPIIC19; CYPIIC8; CYPIIC9; MP-12/MP-20; P450-6B/29C; P450-J; P450C2C; P450C2E; P450IIC17; P450IIC19; P450IIC9
UniProt ID: (Human) P33260, (Human) P33261, (Human) P10632, (Human) P11712, (Human) P05181, (Mouse) Q05421
Entrez Gene ID: (Human) 1562, (Human) 1557, (Human) 1558, (Human) 1559, (Human) 1571, (Mouse) 13106
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support