Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Recommended positive controls: HeLa, mouse lung.
Predicted reactivity: Mouse (92%), Rat (92%), Xenopus laevis (84%), Dog (91%), Pig (92%), Rabbit (92%), Chicken (85%), Rhesus Monkey (99%), Bovine (92%), Guinea pig (91%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an endoplasmic reticulum membrane oxidoreductase with an FAD-binding domain and a flavodoxin-like domain. The protein binds two cofactors, FAD and FMN, which allow it to donate electrons directly from NADPH to all microsomal P450 enzymes. Mutations in this gene have been associated with various diseases, including apparent combined P450C17 and P450C21 deficiency, amenorrhea and disordered steroidogenesis, congenital adrenal hyperplasia and Antley-Bixler syndrome.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCR; CPR; CY DKFZp686G04235; DKFZp686G04235; FLJ26468; NADPH Cytochrome; NADPH--cytochrome P450 reductase; NADPH-dependent cytochrome P450 reductase; P450 (cytochrome) oxidoreductase; P450 Reductase
Gene Aliases: 4933424M13Rik; CPR; CYPOR; P450R; POR
UniProt ID: (Human) P16435, (Mouse) P37040
Entrez Gene ID: (Human) 5447, (Mouse) 18984
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support