Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: MEKVPGEME IERRERSEEL SEAERKAVQA MWARLYASCE DVGVAILVRF FVNFPSAKQY FSQFKHMEDP LEMERSPQLR KHACRVMGAL NTVVENLHDP DKVSSVLALV GKAHALKHKV EPVYFKILSG VILEVVAEEF ASDFPPETQR AWAKLRGLIY SHVTAAYKEV GWVQQVPNAT TPPATLPSSG P (1-190 aa encoded by BC029798)
Cytoglobin is a ubiquitously expressed hexacoordinate hemoglobin that may facilitate diffusion of oxygen through tissues, scavenge nitric oxide or other reactive oxygen species, or serve a protective function during oxidative stress (Trent and Hargrove, 2002).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytoglobin; HGb; Histoglobin; Nitric oxygen dioxygenase; Nitric oxygen dioxygenase CYGB; Nitrite reductase CYGB; NOD; Pseudoperoxidase CYGB; stellate cell activation associated protein; Stellate cell activation-associated protein; Superoxide dismutase CYGB
Gene Aliases: 3110001K20Rik; CYGB; HGB; Staap; STAP
UniProt ID: (Human) Q8WWM9, (Mouse) Q9CX80, (Rat) Q921A4
Entrez Gene ID: (Pig) 100518861, (Human) 114757, (Mouse) 114886, (Rat) 170520
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support