Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 6
MA1-062 detects recombinant human and mouse cytohesin-1 and 2 (ARNO).
MA1-062 has been successfully used in Western blot, IF and immunoprecipitation procedures. By Western blot, this antibody detects an ~48 kDa band representing both cytohesin-1 and cytohesin-2. In immunoprecipitation procedures MA1-062 only precipitates cytohesin-2.
The MA1-062 immunogen is purified, His-tagged cytohesin-1.
The protein encoded by this gene is a member of the PSCD family. Members of this family have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology (PH) domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein (GEP) activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. This gene is highly expressed in natural killer and peripheral T cells, and regulates the adhesiveness of integrins at the plasma membrane of lymphocytes. The encoded protein is 83% homologous to that of CYTH2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARF exchange factor; ARF nucleotide-binding site opener; B2-1/ARNO; CLM1; CLM2; CTC-273B12.8; cytoadhesin 1; Cytohesin-1; Cytohesin-2; homolog of secretory protein SEC7; mSec7-1; mSec7-2; PH, SEC7 and coiled-coil domain-containing protein 1; PH, SEC7 and coiled-coil domain-containing protein 2; pleckstrin homology, Sec7 and coiled-coil domains 1; pleckstrin homology, Sec7 and coiled-coil domains 2; pleckstrin homology, Sec7 and coiled-coil domains 2 (cytohesin-2); pleckstrin homology, Sec7 and coiled/coil domains 2 (cytohesin-2); Protein ARNO; SEC7 homolog A; SEC7 homolog B; SEC7 homolog B2-1
Gene Aliases: ARNO; B2-1; CLM1; CLM2; CTH-1; CTS18; CTS18.1; CYTH1; CYTH2; CYTIP; CYTOHESIN-1; D17S811E; mKIAA4240; PSCD1; PSCD2; PSCD2L; SEC7; Sec7b; SEC7L; Sec7p-L; Sec7p-like
UniProt ID: (Human) Q15438, (Human) Q99418, (Mouse) Q9QX11, (Mouse) P63034
Entrez Gene ID: (Human) 9267, (Human) 9266, (Mouse) 19157, (Mouse) 19158
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support