Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 5
Positive test controls include: Rat eye.
Immunogen sequence: EMQIESLNEE LAYMKKNHED ELQSFRVGGP GEVSVEMDAA PGVDLTRLLN DMRAQYETIA EQNRKDAEAW FIEKSGELRK EISTNTEQLQ SSKSEVTDLR RAFQNLEIEL QSQLAMKKSL EDSLAEAEGD YCAQLSQVQQ LISNLEAQLL QVRADAERQN VDHQRLLNVK ARLELEIETY RRLLDGEAQG DGLEESLFVT DSKSQAQSTD SSKDPTKTRK IKTVVQEMVN GEVVSSQVQE IEELM
KRT12 encodes the type I intermediate filament chain keratin 12, expressed in corneal epithelia. Mutations in this gene lead to Meesmann corneal dystrophy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-12; cytokeratin-12; K12; keratin 12 (Meesmann corneal dystrophy); keratin complex 1, acidic, gene 12; keratin K12; keratin, type I cytoskeletal 12; Keratin, type I cytoskeletal 12 (Cytokeratin 12); keratin-12; krt12 {ECO:0000250|UniProtKB:Q99456}; type I keratin KA12
Gene Aliases: AI835216; K12; Ka12; Krt-12; Krt1-12
Entrez Gene ID: (Mouse) 268482, (Rat) 360625
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support