Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Positive Control: Mouse eyeball tissue lysates, rat eyeball tissue.
Subcellular Location: Intermediate filament, cytosol, extracellular exosome.
KRT12 encodes the type I intermediate filament chain keratin 12, expressed in corneal epithelia. Mutations in this gene lead to Meesmann corneal dystrophy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-12; Cytokeratin-12; K12; keratin 12 (Meesmann corneal dystrophy); keratin 12, type I; keratin complex 1, acidic, gene 12; keratin K12; Keratin, type I cytoskeletal 12; Keratin, type I cytoskeletal 12 (Cytokeratin 12); Keratin-12; krt12 {ECO:0000250|UniProtKB:Q99456}; type I keratin KA12
Gene Aliases: AI835216; K12; Ka12; Krt-12; Krt1-12; KRT12
UniProt ID: (Human) Q99456
Entrez Gene ID: (Human) 3859, (Mouse) 268482, (Rat) 360625
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support