Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Positive Samples: Cal-27
Immunogen sequence: YATQLQQIQG LIGGLEAQLS ELRCEMEAQN QEYKMLLDIK TRLEQEIATY RSLLEGQDAK MAGIAIREAS SGGGGSSSNF HINVEESVDG QVVSSHKREI
Cytokeratin 15 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region on chromosome 17q21. 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C K1CO; CK-15; cytokeratin 15; Cytokeratin-15; K15; keratin 15, type I; Keratin, type I cytoskeletal 15; Keratin-15; keratin-15, basic; keratin-15, beta; KRT1; krt15 {ECO:0000250|UniProtKB:P19012}; type I cytoskeletal 15; Type I keratin Ka15
Gene Aliases: CK15; K15; K1CO; Ka15; KRT15; KRTB
UniProt ID: (Human) P19012, (Rat) Q6IFV3
Entrez Gene ID: (Human) 3866, (Rat) 287700
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support