Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Cytokeratin 17 Antibodies

            Invitrogen

            Cytokeratin 17 Monoclonal Antibody (A2B9)

            View all (54) Cytokeratin 17 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # MA5-42398
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            494.00
            100 µL
             
            Estimated availability date: 09-Jun-2025
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite Cytokeratin 17 Monoclonal Antibody (A2B9)

            • Antibody Testing Data (5)
            Application
            Cytokeratin 17 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 5

            Cytokeratin 17 Antibody (MA5-42398) in IHC (P)

            Immunohistochemistry (Paraffin) analysis of paraffin-embedded human liver carcinoma tissue using Cytokeratin 17 Monoclonal Antibody (Product # MA5-42398). The section was pre-treated using heat mediated antigen retrieval with Tris-EDTA buffer (pH 8.0-8.4) for 20 minutes. The tissues were blocked in 5% BSA for 30 minutes at room temperature, washed with ddH2O and PBS, and then probed with the Cytokeratin 17 ... View More
            View Product

            Cytokeratin 17 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Cytokeratin 17 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Cytokeratin 17 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Cytokeratin 17 Antibody in Western Blot (WB)
            Cytokeratin 17 Antibody in Flow Cytometry (Flow)

            Product Details

            MA5-42398

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:2,000
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            1:50-1:200
            -

            Flow Cytometry (Flow)

            1:50-1:100
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Mouse / IgG1

            Class

            Monoclonal

            Type

            Antibody

            Clone

            A2B9

            Immunogen

            Synthetic peptide within Human Cytokeratin 17 aa 383-432/432
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            2 mg/mL

            Purification

            Protein G

            Storage buffer

            PBS, pH 7.4, with 50% glycerol, 0.2% BSA

            Contains

            0.05% sodium azide

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2911539

            Product Specific Information

            Positive Control: SiHa cell lysate, human tonsil tissue, human liver carcinoma tissue, human skin tissue.

            Subcellular Location: Cytoplasm.

            Target Information

            Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between approximately40 kDa and approximately68 kDa and isoelectric pH between 4. 9 - 7. 8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: 39.1; CK-17; CK17; Cytokeratin-17; hard keratin; K17; keratin 17, type I; Keratin, type I cytoskeletal 17; Keratin-17

            View more View less

            Gene Aliases: 39.1; CK-17; K17; KRT17; PC; PC2; PCHC1

            View more View less

            UniProt ID: (Human) Q04695

            View more View less

            Entrez Gene ID: (Human) 3872

            View more View less

            Function(s)

            structural constituent of cytoskeleton protein binding MHC class II receptor activity MHC class II protein binding

            Process(es)

            signal transduction epidermis development positive regulation of cell growth hair follicle morphogenesis keratinization intermediate filament organization positive regulation of translation positive regulation of hair follicle development
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-x5c57:80/100.66.78.18:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0