Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: MQNLNDRLA SYLEKVRTLE QSNSKLEVQI KQWYETNAPR AGRDYSAYYR QIEELRSQIK DAQLQNARCV LQIDNAKLAA EDFRLKYETE RGIRLTVEAD LQGLNKVFDD LTLHKTDLEI QIEELNKDLA LLKKEHQEEV DGLHKHLGNT VNVEVDAAPG LNLGVIMNEM RQKYEVMAQK NLQEAKEQFE RQTAVLQQQV TVNTEELKGT EVQLTELRRT SQSLEIELQS HLSMKESLEH TLEETKARYS SQLANLQSLL SSLEAQLMQI RSNMERQNNE YHILLDIKTR LEQEIATYRR LLEGEDVKTT EYQLSTLEER DIKKTRKIKT VVQEVVDGKV VSSEVKEVEE NI (74-424 aa encoded by BC031559)
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-20; CK-21; cytokeratin 20; cytokeratin 21; Cytokeratin-20; Cytokeratin-21; K20; keratin 20, type I; Keratin, type I cytoskeletal 20; Keratin-20; krt20; MGC35423; Protein IT
Gene Aliases: 9030623C06Rik; CD20; CK-20; CK20; K20; KRT20; KRT21
UniProt ID: (Human) P35900, (Mouse) Q9D312, (Rat) P25030
Entrez Gene ID: (Human) 54474, (Mouse) 66809, (Rat) 286912
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support