Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute lyophilized product with 1 mL distilled water.
IT-Ks 20.8 represents an excellent marker for certain types of carcinomas such as adenocarcinomas of the colon, transitional cell carcinomas of the bladder and Merkel cell tumors of the skin. Very sensitive detection of intestinal and gastric foveolar epithelium, urothelial umbrella cells, Merkel cells of epidermis as well as tumors originating therefrom (e.g. primary and metastatic colorectal carcinoma). Adenocarcinomas of breast, lung, endometrium and ovary (non-mucinous) as well as neuroendocrine tumors of the lung are essentially negative. Polypeptide recognized: protein IT (keratin K20; MW 46 000; formerly also designated cytokeratin 20).Tested cultured cell lines: RT-4.
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-20; cytokeratin 20; Cytokeratin-20; K20; keratin 20, type I; Keratin, type I cytoskeletal 20; Keratin-20; krt20; MGC35423; Protein IT
Gene Aliases: 9030623C06Rik; CD20; CK-20; CK20; K20; KRT20; KRT21
UniProt ID: (Human) P35900, (Mouse) Q9D312
Entrez Gene ID: (Human) 54474, (Mouse) 66809
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support