Product References
Application of immune enhanced organoids in modeling personalized Merkel cell carcinoma research.
Scientific reports
Forsythe SD,Erali RA,Laney P,Sivakumar H,Li W,Skardal A,Soker S,Votanopoulos KI
MA5-13263 was used in Immunohistochemistry to study the application of patient tumor organoids (PTOs) in modelling personalized research in Merkel cell carcinoma (MCC).
Tue Aug 16 00:00:00 UTC 2022
In vivo and in vitro cell-based model of lung adenocarcinoma from patients with pleural effusion.
Neoplasma
Popovic M,Liu Y,Lattova E,Mann D,Curreli S,Zdráhal Z,Edelman M,Bryant J
MA5-13263 was used in Immunohistochemistry to enable broader experimental approaches in studies of lung adenocarcinoma.
Sat May 01 00:00:00 UTC 2021
Organoid Platform in Preclinical Investigation of Personalized Immunotherapy Efficacy in Appendiceal Cancer: Feasibility Study.
Clinical cancer research : an official journal of the American Association for Cancer Research
Forsythe SD,Erali RA,Sasikumar S,Laney P,Shelkey E,D'Agostino R,Miller LD,Shen P,Levine EA,Soker S,Votanopoulos KI
MA5-13263 was used in Immunohistochemistry to investigate preclinical study for the application of immunotherapy in treating appendiceal cancer using a personalized organoid model to demonstrate that immunotherapy shows measurable efficacy.
Wed Sep 15 00:00:00 UTC 2021
Antibody targeting of B7-H4 enhances the immune response in urothelial carcinoma.
Oncoimmunology
Podojil JR,Glaser AP,Baker D,Courtois ET,Fantini D,Yu Y,Eaton V,Sivajothi S,Chiang M,Das A,McLaughlin KA,Robson P,Miller SD,Meeks JJ
MA5-13263 was used in Immunohistochemistry-immunofluorescence to suggest that antibodies targeting B7-H4 may be a viable strategy for bladder cancers unresponsive to PD-1 checkpoint inhibitors.
Wed Jul 28 00:00:00 UTC 2021
Antibody targeting of B7-H4 enhances the immune response in urothelial carcinoma.
Oncoimmunology
Podojil JR,Glaser AP,Baker D,Courtois ET,Fantini D,Yu Y,Eaton V,Sivajothi S,Chiang M,Das A,McLaughlin KA,Robson P,Miller SD,Meeks JJ
MA5-13263 was used in Immunohistochemistry-immunofluorescence to suggest that antibodies targeting B7-H4 may be a viable strategy for bladder cancers unresponsive to PD-1 checkpoint inhibitors.
Wed Jul 28 00:00:00 UTC 2021
Electroporation with Cisplatin against Metastatic Pancreatic Cancer: In Vitro Study on Human Primary Cell Culture.
BioMed research international
Michel O,Kulbacka J,Saczko J,Mączyńska J,Błasiak P,Rossowska J,Rzechonek A
MA5-13263 was used in Immunocytochemistry to confirm the importance of primary cells models in studies on the efficacy of experimental cancer therapies.
Fri Sep 28 00:00:00 UTC 2018
Mutant p53 cancers reprogram macrophages to tumor supporting macrophages via exosomal miR-1246.
Nature communications
Cooks T,Pateras IS,Jenkins LM,Patel KM,Robles AI,Morris J,Forshew T,Appella E,Gorgoulis VG,Harris CC
MA5-13263 was used in Immunohistochemistry to support a microenvironmental gain of oncogenic functions role for mutant-p53 in engaging the immune system to promote cancer progression and metastasis.
Thu Feb 22 00:00:00 UTC 2018
Complex APC germline mutation associated metaplasia and intraepithelial neoplasia (CAM-IEN) of the gallbladder.
Pathology, research and practice
Böger C,Haag J,Egberts JH,Röcken C
MA5-13263 was used in immunohistochemistry - paraffin section to report a unique case of familial adenomatous polyposis
Fri Jan 01 00:00:00 UTC 2016
Ectopic Atoh1 expression drives Merkel cell production in embryonic, postnatal and adult mouse epidermis.
Development (Cambridge, England)
Ostrowski SM,Wright MC,Bolock AM,Geng X,Maricich SM
MA5-13263 was used in immunohistochemistry - free floating to analyze the drive of Merkel cell production in postnatal, embryonic and adult mouse epidermis by ectopic Atoh1 expression
Wed Jul 15 00:00:00 UTC 2015
Increased utilization, verification, and clinical implications of immunocytochemistry: Experience in a northern New England hospital.
Diagnostic cytopathology
Sauter JL,Ambaye AB,Mount SL
MA5-13263 was used in immunohistochemistry to study a northern New England hospital and increased utilization, verification, and clinical implications of immunocytochemistry
Tue Sep 01 00:00:00 UTC 2015
Adenocarcinoma with sarcomatous dedifferentiation arising from mature cystic teratoma of the anterior mediastinum.
Pathology, research and practice
Kim JY,Lee CH,Park WY,Kim JY,Kim AR,Shin N,Park DY,Huh GY
MA5-13263 was used in immunohistochemistry to report on a case of adenocarcinoma with sarcomatous dedifferentiation arising from mature cystic teratoma of the anterior mediastinum
Sat Dec 15 00:00:00 UTC 2012
Miliary brain metastases from occult lung adenocarcinoma: Radiologic and histopathologic confirmation.
Journal of neurosciences in rural practice
Kahveci R,Gürer B,Kaygusuz G,Sekerci Z
MA5-13263 was used in immunohistochemistry to report on a case of miliary brain metastases from occult lung adenocarcinoma
Sat Sep 01 00:00:00 UTC 2012
Primary mucinous cystadenoma of the spermatic cord within the inguinal canal.
Diagnostic pathology
Kim JY,Lee YT,Kang HJ,Lee CH
MA5-13263 was used in immunohistochemistry to report on a case of primary mucinous cystadenoma of the spermatic cord
Mon Oct 08 00:00:00 UTC 2012
Huntingtin-interacting protein 1: a Merkel cell carcinoma marker that interacts with c-Kit.
The Journal of investigative dermatology
Ames HM,Bichakjian CK,Liu GY,Oravecz-Wilson KI,Fullen DR,Verhaegen ME,Johnson TM,Dlugosz AA,Ross TS
MA5-13263 was used in immunohistochemistry to evaluate Huntingtin-interacting protein 1 as a marker for Merkel cell carcinoma
Sat Oct 01 00:00:00 UTC 2011
The presence of Merkel cells and CD10- and CD34-positive stromal cells compared in benign and malignant oral tumors.
Oral diseases
Gao HW,Ho JY,Lee HS,Yu CP
MA5-13263 was used in immunohistochemistry to study the role of Merkel cells and CD10(+) and CD34(+) stromal cells in the transition from benign to malignant oral lesions
Fri May 01 00:00:00 UTC 2009
Tumourigenic effect of Schistosoma haematobium total antigen in mammalian cells.
International journal of experimental pathology
Botelho M,Oliveira P,Gomes J,Gartner F,Lopes C,da Costa JM,Machado JC
MA5-13263 was used in immunohistochemistry to investigate the influence of S. haematobium total antigen on tumor development
Sat Aug 01 00:00:00 UTC 2009
Giant cell tumor-like lesion of the urinary bladder: a report of two cases and literature review; giant cell tumor or undifferentiated carcinoma?
Diagnostic pathology
Behzatoğlu K,Durak H,Canberk S,Aydin O,Huq GE,Oznur M,Ozyalvaçli G,Yildiz P
MA5-13263 was used in immunohistochemistry to report on two cases of giant cell tumor-like lesion of the urinary bladder
Thu Dec 31 00:00:00 UTC 2009
Keratin profiling in the developing human prostate. A different approach to understanding epithelial lineage.
Anticancer research
Trompetter M,Smedts F,van der Wijk J,Schoots C,de Jong HJ,Hopman A,de la Rosette J
MA5-13263 was used in immunohistochemistry to investigate the distribution of keratin in prostate during development
Thu Apr 10 00:00:00 UTC 2008
Mucoepidermoid carcinoma of the parotid gland with ovarian and peritoneal metastasis.
The journal of obstetrics and gynaecology research
Buyukkurt S,Altintas A,Gumurdulu D,Zeren H,Guzel B
MA5-13263 was used in immunohistochemistry to investigate the metastasis of mucoepidermoid carcinoma of the parotid gland
Tue Apr 01 00:00:00 UTC 2008
Basaloid adenocarcinoma. A new variant of pulmonary adenocarcinoma.
Virchows Archiv : an international journal of pathology
Marci V,Volante M,Cappia S,Righi L,Novello C,Scagliotti GV,Brambilla E,Papotti M
MA5-13263 was used in immunohistochemistry to report a clinical case of basaloid adenocarcinoma
Sat Sep 01 00:00:00 UTC 2007
Combination chemotherapy with gemcitabine and cisplatin as first-line treatment for immunohistochemically proven cholangiocarcinoma.
American journal of clinical oncology
Lee GW,Kang JH,Kim HG,Lee JS,Lee JS,Jang JS
MA5-13263 was used in immunohistochemistry to investigate the combination chemotherapy of gemcitabine and cisplatin against cholangiocarcinoma
Sat Apr 01 00:00:00 UTC 2006
Reconstitution of human telomerase reverse transcriptase expression rescues colorectal carcinoma cells from in vitro senescence: evidence against immortality as a constitutive trait of tumor cells.
Cancer research
Dalerba P,Guiducci C,Poliani PL,Cifola I,Parenza M,Frattini M,Gallino G,Carnevali I,Di Giulio I,Andreola S,Lombardo C,Rivoltini L,Schweighoffer T,Belli F,Colombo MP,Parmiani G,Castelli C
MA5-13263 was used in immunocytochemistry to study the rescue of colorectal cancer cells from senescence by the reconstitution of telomerase reverse transcriptase activity
Tue Mar 15 00:00:00 UTC 2005
Mesenchymal hamartoma of the liver in adulthood: immunohistochemical profiles, clinical and histopathological features in two patients.
Journal of hepato-biliary-pancreatic surgery
Yesim G,Gupse T,Zafer U,Ahmet A
MA5-13263 was used in immunohistochemistry to study two adult patients with mesenchymal hamartoma of the liver
Thu Jul 06 00:00:00 UTC 2006
CDX-2 homeobox gene expression is a reliable marker of colorectal adenocarcinoma metastases to the lungs.
The American journal of surgical pathology
Barbareschi M,Murer B,Colby TV,Chilosi M,Macri E,Loda M,Doglioni C
MA5-13263 was used in immunohistochemistry to investigate the diagnostic value of CDX-2 in lung metastases from colorectal carcinomas
Sat Feb 01 00:00:00 UTC 2003