Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Recommended positive controls: A549, HeLa.
Predicted reactivity: Mouse (89%), Rat (87%), Pig (87%), Chimpanzee (100%), Bovine (86%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hair keratin; Hair keratin, type I Ha1; hard keratin, type I, 1; K31; keratin 31, type I; keratin, hair, acidic,1; Keratin, type I cuticular Ha1; Keratin-31; KRT hHa1
Gene Aliases: Ha-1; HA1; HHA1; HKA1; KRT31; KRTHA1
UniProt ID: (Human) Q15323
Entrez Gene ID: (Human) 3881
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support