Product References
The Isolation and In Vitro Differentiation of Primary Fetal Baboon Tracheal Epithelial Cells for the Study of SARS-CoV-2 Host-Virus Interactions.
Viruses
Subramaniyan B,Gurung S,Bodas M,Moore AR,Larabee JL,Reuter D,Georgescu C,Wren JD,Myers DA,Papin JF,Walters MS
PA1-37974 was used in Immunohistochemistry to develop an in vitro NHP model that provides a platform for the study of SARS-CoV-2 infection and other human respiratory viruses.
Tue Mar 28 00:00:00 UTC 2023
MIR503HG silencing promotes endometrial stromal cell progression and metastasis and suppresses apoptosis in adenomyosis by activating the Wnt/β‑catenin pathway via targeting miR‑191.
Experimental and therapeutic medicine
Xu X,Cai B,Liu Y,Liu R,Li J
PA5-32465 was used in Immunocytochemistry to investigate the specific role of MIR503HG in adenomyosis pathogenesis and progression using endometrial stromal cells derived from the endometrium of patients with adenomyosis as a model.
Wed Mar 01 00:00:00 UTC 2023
Assessment of Beta-2 Microglobulin Gene Edited Airway Epithelial Stem Cells as a treatment for Sulfur Mustard Inhalation.
Frontiers in genome editing
Naeimi Kararoudi M,Alsudayri A,Hill CL,Elmas E,Sezgin Y,Thakkar A,Hester ME,Malleske DT,Lee DA,Neal ML,Perry MR,Harvilchuck JA,Reynolds SD
PA5-32465 was used in Immunohistochemistry to generate B2M-knockout tracheobronchial airway epithelial stem cell , which are termed Universal Donor Stem Cells (UDSC), and characterise them.
Fri Feb 25 00:00:00 UTC 2022
A simple method to generate human airway epithelial organoids with externally orientated apical membranes.
American journal of physiology. Lung cellular and molecular physiology
Boecking CA,Walentek P,Zlock LT,Sun DI,Wolters PJ,Ishikawa H,Jin BJ,Haggie PM,Marshall WF,Verkman AS,Finkbeiner WE
PA1-37974 was used in Immunocytochemistry-immunofluorescence to describe a simple method to generate organoid cultures using isolated human tracheobronchial epithelial cells grown in mixed matrix components and supplemented at day 14 with the Wnt pathway agonist R-spondin 2 (RSPO2) and the bone morphogenic protein antagonist Noggin.
Tue Mar 01 00:00:00 UTC 2022
Cigarette Smoke Activates NOTCH3 to Promote Goblet Cell Differentiation in Human Airway Epithelial Cells.
American journal of respiratory cell and molecular biology
Bodas M,Moore AR,Subramaniyan B,Georgescu C,Wren JD,Freeman WM,Brown BR,Metcalf JP,Walters MS
PA1-37974 was used in Immunohistochemistry (Paraffin) to identify NOTCH3 activation as one of the important mechanisms by which cigarette smoke induces goblet cell differentiation, thus providing a novel potential strategy to control GCMH-related pathologies in smokers and patients with COPD.
Thu Apr 01 00:00:00 UTC 2021
The NOTCH3 Downstream Target HEYL Is Required for Efficient Human Airway Basal Cell Differentiation.
Cells
Bodas M,Subramaniyan B,Moore AR,Metcalf JP,Ocañas SR,Freeman WM,Georgescu C,Wren JD,Walters MS
PA1-37974 was used in Immunocytochemistry to investigate the mechanisms of NOTCH3 receptor signaling regulating basal cells to club cell differentiation.
Thu Nov 18 00:00:00 UTC 2021
Role of KRAS in regulating normal human airway basal cell differentiation.
Respiratory research
Ogawa F,Walters MS,Shafquat A,O'Beirne SL,Kaner RJ,Mezey JG,Zhang H,Leopold PL,Crystal RG
Published figure using Cytokeratin 5 polyclonal antibody (Product # PA5-32465) in Immunofluorescence
Fri Aug 09 00:00:00 UTC 2019
Role of KRAS in regulating normal human airway basal cell differentiation.
Respiratory research
Ogawa F,Walters MS,Shafquat A,O'Beirne SL,Kaner RJ,Mezey JG,Zhang H,Leopold PL,Crystal RG
Published figure using Cytokeratin 5 polyclonal antibody (Product # PA5-32465) in Immunofluorescence
Fri Aug 09 00:00:00 UTC 2019
ΔN-Tp63 Mediates Wnt/β-Catenin-Induced Inhibition of Differentiation in Basal Stem Cells of Mucociliary Epithelia.
Cell reports
Haas M,Gómez Vázquez JL,Sun DI,Tran HT,Brislinger M,Tasca A,Shomroni O,Vleminckx K,Walentek P
Published figure using Cytokeratin 5 polyclonal antibody (Product # PA5-32465) in Immunocytochemistry
Tue Sep 24 00:00:00 UTC 2019
Characterization of an immortalized human small airway basal stem/progenitor cell line with airway region-specific differentiation capacity.
Respiratory research
Wang G,Lou HH,Salit J,Leopold PL,Driscoll S,Schymeinsky J,Quast K,Visvanathan S,Fine JS,Thomas MJ,Crystal RG
PA5-32465 was used in Immunocytochemistry-immunoflourescence to generate and characterise an immortalized human small airway basal cell line.
Fri Aug 23 00:00:00 UTC 2019
Na+/H+ Exchangers Are Required for the Development and Function of Vertebrate Mucociliary Epithelia.
Cells, tissues, organs
Sun DI,Tasca A,Haas M,Baltazar G,Harland RM,Finkbeiner WE,Walentek P
PA1-37974 was used in Immunohistochemistry to study the contribution of Na+/H+ exchangers to signalling, development and pathogenesis in the human respiratory tract.
Mon Aug 26 00:00:00 UTC 2019
Mandatory role of HMGA1 in human airway epithelial normal differentiation and post-injury regeneration.
Oncotarget
Zhang H,Yang J,Walters MS,Staudt MR,Strulovici-Barel Y,Salit J,Mezey JG,Leopold PL,Crystal RG
Tue Mar 06 00:00:00 UTC 2018
Endothelial Cell Mediated Promotion of Ciliated Cell Differentiation of Human Airway Basal Cells via Insulin and Insulin-Like Growth Factor 1 Receptor Mediated Signaling.
Stem cell reviews and reports
Gomi K,Tang Y,Arbelaez V,Crystal RG,Walters MS
PA137974 was used in immunocytochemistry to attempt to understand the role of human airway basal cells and endothelial cells cross-talk in regulating basal cell stem/progenitor function
Sat Apr 01 00:00:00 UTC 2017
Endothelial Cell Mediated Promotion of Ciliated Cell Differentiation of Human Airway Basal Cells via Insulin and Insulin-Like Growth Factor 1 Receptor Mediated Signaling.
Stem cell reviews and reports
Gomi K,Tang Y,Arbelaez V,Crystal RG,Walters MS
PA137974 was used in immunocytochemistry to attempt to understand the role of human airway basal cells and endothelial cells cross-talk in regulating basal cell stem/progenitor function
Sat Apr 01 00:00:00 UTC 2017
JAG1-Mediated Notch Signaling Regulates Secretory Cell Differentiation of the Human Airway Epithelium.
Stem cell reviews and reports
Gomi K,Staudt MR,Salit J,Kaner RJ,Heldrich J,Rogalski AM,Arbelaez V,Crystal RG,Walters MS
PA137974 was used in immunocytochemistry to determine the role of JAG1 human basal cell differentiation
Mon Aug 01 00:00:00 UTC 2016
Endothelial MMP14 is required for endothelial-dependent growth support of human airway basal cells.
Journal of cell science
Ding BS,Gomi K,Rafii S,Crystal RG,Walters MS
PA1-37974 was used in immunocytochemistry to investigate factors involved in the crosstalk between basal cells and endothelial cells.
Sat Aug 15 00:00:00 UTC 2015
Activation of NOTCH1 or NOTCH3 signaling skews human airway basal cell differentiation toward a secretory pathway.
PloS one
Gomi K,Arbelaez V,Crystal RG,Walters MS
PA1-37974 was used in immunocytochemistry and western blot to investigate the role of Notch signaling in human basal cell differentiation
Wed Jan 13 00:00:00 UTC 2016
Activation of soluble guanylyl cyclase prevents foam cell formation and atherosclerosis.
Acta physiologica (Oxford, England)
Tsou CY,Chen CY,Zhao JF,Su KH,Lee HT,Lin SJ,Shyue SK,Hsiao SH,Lee TS
PA1-37974 was used in western blot to study the mechanism by which soluble guanylyl cyclase activation blocks macrophage foam cell formation and atherosclerosis
Tue Apr 01 00:00:00 UTC 2014
Generation of a human airway epithelium derived basal cell line with multipotent differentiation capacity.
Respiratory research
Walters MS,Gomi K,Ashbridge B,Moore MA,Arbelaez V,Heldrich J,Ding BS,Rafii S,Staudt MR,Crystal RG
PA1-37974 was used in western blot to generate an immortalized human epithelial basal cell line capable of multipotent differentiation
Tue Dec 03 00:00:00 UTC 2013
Generation of a human airway epithelium derived basal cell line with multipotent differentiation capacity.
Respiratory research
Walters MS,Gomi K,Ashbridge B,Moore MA,Arbelaez V,Heldrich J,Ding BS,Rafii S,Staudt MR,Crystal RG
PA1-37974 was used in western blot to generate an immortalized human epithelial basal cell line capable of multipotent differentiation
Tue Dec 03 00:00:00 UTC 2013