Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Cytokeratin 5 Antibodies

            Invitrogen

            Cytokeratin 5 Monoclonal Antibody (XM26)

            10 Published Figures
            47 References
            View all (81) Cytokeratin 5 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # MA5-12596
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            487.00
            500 µL
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite Cytokeratin 5 Monoclonal Antibody (XM26)

            • Antibody Testing Data (8)
            • Published Figures (10)
            Application
            Cytokeratin 5 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 18

            Cytokeratin 5 Antibody (MA5-12596) in ICC/IF

            Immunofluorescence analysis of Cytokeratin 5 was performed using 70% confluent log phase A-431 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with Cytokeratin 5 (XM26) Mouse Monoclonal Antibody (Product # MA5-12596) at 1:250 dilution in 0.1% BSA and incub... View More
            View Product

            Cytokeratin 5 Antibody in Immunocytochemistry (ICC/IF)
            Cytokeratin 5 Antibody in Immunocytochemistry (ICC/IF)
            Cytokeratin 5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Cytokeratin 5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Cytokeratin 5 Antibody in Immunohistochemistry (Frozen) (IHC (F))
            Cytokeratin 5 Antibody in Western Blot (WB)
            Cytokeratin 5 Antibody in Western Blot (WB)
            Cytokeratin 5 Antibody in Western Blot (WB)
            Cytokeratin 5 Antibody in Immunocytochemistry (ICC/IF)
            Cytokeratin 5 Antibody in Immunocytochemistry, Immunohistochemistry (Paraffin) (ICC/IF, IHC (P))
            Cytokeratin 5 Antibody in Immunocytochemistry (ICC/IF)
            Cytokeratin 5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Cytokeratin 5 Antibody in Immunohistochemistry (IHC)
            Cytokeratin 5 Antibody in Immunohistochemistry (IHC)
            Cytokeratin 5 Antibody in Immunohistochemistry (IHC)
            Cytokeratin 5 Antibody in Immunohistochemistry (IHC)
            Cytokeratin 5 Antibody in Western Blot (WB)
            Cytokeratin 5 Antibody in Western Blot (WB)

            Product Details

            MA5-12596

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:1,000
            View 2 publications 2 publications

            Immunohistochemistry (IHC)

            -
            View 29 publications 29 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:20-1:100
            View 6 publications 6 publications

            Immunohistochemistry (Frozen) (IHC (F))

            1:2,000
            -

            Immunocytochemistry (ICC/IF)

            1:200-1:250
            View 8 publications 8 publications

            Immunoprecipitation (IP)

            -
            View 1 publication 1 publication

            In vitro Assay (IV)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human

            Published species

            Human, Mouse

            Host/Isotype

            Mouse / IgG1, kappa

            Class

            Monoclonal

            Type

            Antibody

            Clone

            XM26

            Immunogen

            Recombinant protein corresponding to C-terminal 103 aa of cytokeratin 5
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            Conc. Not Determined

            Storage buffer

            tissue culture supernatant

            Contains

            0.09% sodium azide

            Storage conditions

            4°C

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_11008032

            Product Specific Information

            MA5-12596 targets Cytokeratin 5 in IHC (P) applications and shows reactivity with Human samples.

            The MA5-12596 immunogen is recombinant protein corresponding to C-terminal 103 aa of cytokeratin 5.

            Target Information

            Cytokeratin pan is part of a subfamily of intermediate filament proteins that are characterized by remarkable biochemical diversity, and represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight between 40 kDa- 68 kDa, and an isoelectric pH between 4.9-7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. The composition of cytokeratin pairs vary with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: 58 kDa cytokeratin; CK-5; Cytokeratin-5; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; K5; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; Keratin, type II cytoskeletal 5; Keratin-5; Type-II keratin Kb5

            View more View less

            Gene Aliases: CK5; DDD; DDD1; EBS2; K5; KRT5; KRT5A

            View more View less

            UniProt ID: (Human) P13647

            View more View less

            Entrez Gene ID: (Human) 3852

            View more View less

            Function(s)

            structural constituent of cytoskeleton protein binding scaffold protein binding

            Process(es)

            cytoskeleton organization epidermis development hemidesmosome assembly
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-f2ctl:80/100.66.76.208:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0