Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: FEQYINNLR RQLDNIVGER GRLDSELRNM QDLVEDLKNK YEDEINKRTA AENEFVTLKK DVDAAYMNKV ELQAKADTLT DEINFLRALY DAELSQMQTH ISDTSVVLSM DNNRNLDLDS IIAEVKAQYE EIAQRSRAEA ESWYQTKYEE LQITAGRHGD DLRNTKQEIA EINRMIQRLR SEIDHVKKQC ANLQAAIADA EQRGEMALKD AKNKLEGLED ALQKAKQDLA RLLKEYQELM NVKLALDVEI ATYRKLLEGE ECRLNGEGVG QVNISVVQST VSSGYGGASG VGSGLGLGGG SSYSYGSGLG VGGGFSSSSG RATGGGLSSV GGGSSTIKYT TTSSSSRKSY KH (214-564 aa encoded by BC034535)
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. As many as six of this type II cytokeratin (KRT6) have been identified; the multiplicity of the genes is attributed to successive gene duplication events. The genes are expressed with family members KRT16 and/or KRT17 in the filiform papillae of the tongue, the stratified epithelial lining of oral mucosa and esophagus, the outer root sheath of hair follicles, and the glandular epithelia. Mutations in these genes have been associated with pachyonychia congenita. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-6B; cytokeratin 6B; Cytokeratin-6B; K6B; keratin 6B, type II; keratin complex 2, basic, gene 6b; keratin, epidermal, type II, K6B; Keratin, type II cytoskeletal 6B; keratin-6-beta; Keratin-6B; keratin-like 1 (a type II keratin sequence); mK6-beta; Type-II keratin Kb10
Gene Aliases: BB144589; CK-6B; CK6B; K6B; Krt2-6b; KRT6B; KRTL1; mK6[b]; PC2; PC4
UniProt ID: (Human) P04259
Entrez Gene ID: (Human) 3854, (Mouse) 16688
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support