Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
FIGURE: 1 / 1
Immunogen sequence: MSIHFSSPVF TSRSAAFSGR GAQVRLSSAR PGGLGSSSLY GLGASRPRVA VRSAYGGPVG AGIREVTINQ SLLAPLRLDA DPSLQRVRQE ESEQIKTLNN KFASFIDKVR FLEQQNKLLE TKWTLLQEQK SAKSSRLPDI FEAQIAGLRG QLEALQVDGG RLEAELRSMQ DVVEDFKNKY EDEINRRTAA ENEFVVLKKD VDAAYMSKVE LEAKVDALND EINFLRTLNE TELTELQSQI SDTSVVLSMD NSRSLDLDGI IAEVKAQYEE MAKCSRAEAE AWYQTKFETL QAQAGKHGDD LRNTRNEISE MNRAIQRLQA EIDNIKNQRA KLEAAIAEAE ERGELALKDA R
Cytokeratin 7 blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-7; cytokeratin 7; Cytokeratin-7; K7; keratin 7, type II; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; Keratin, type II cytoskeletal 7; Keratin-7; Sarcolectin; type II mesothelial keratin K7; Type-II keratin Kb7
Gene Aliases: CK7; K2C7; K7; KRT7; SCL
UniProt ID: (Human) P08729
Entrez Gene ID: (Human) 3855
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support